Skip to Content
Merck
All Photos(2)

Key Documents

ABN912

Sigma-Aldrich

Anti-Myelin Basic Protein

from rabbit, purified by affinity chromatography

Synonym(s):

MBP

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€153.00
100 μG
€443.00

€153.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€153.00
100 μG
€443.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€153.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

species reactivity (predicted by homology)

guinea pig (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)
rat ... Mbp(24547)

General description

Myelin basic protein (UniProt: also known as MBP) is encoded by the MBP gene in guinea pig. MBP is a homodimeric protein that is found in both the central and the peripheral nervous system. It is one of the most abundant protein component of the myelin membrane in the CNS and plays a role in both the formation and stabilization of this myelin membrane. It is responsible for adhesion of the cytosolic surfaces of multilayered compact myelin. At least 5 charge isomers (C1, C2, C3, C4, and C5) are reported to be produced because of optional post-translational modifications such as phosphorylation of serine or threonine residues, deamidation of glutamine or asparagine residues, citrullination and methylation of arginine residues. Of these C1 isomer is the most cationic, least modified, and most abundant form and C5 is the least cationic form. C1 and C2 are reported to be unphosphorylated, C3 and C4 are monophosphorylated, and C5 is phosphorylated at two positions. Phosphorylation is achieved with the help of TAOK2, VRK2, MAPK11, MAPK12, MAPK14, and MINK1). MBP can interact with many polyanionic proteins including actin, tubulin, calmodulin, and clathrin, and with negatively charged lipids. (Ref.: Boggs, JM (2006). Cell. Mol. Life Sci. 63(17); 1945-61).

Specificity

This rabbit polyclonal antibody detects myelin basic protein. It targets an epitope within 19 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the internal region of Guinea pig myelin basic protein.

Application

Anti-Myelin Basic Protein Antibody, Cat. No. ABN912, is a highly specific rabbit polyclonal antibody that targets Myelin basic protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Myelin Basic Protein in human cerebral cortex and rat cerebellum tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human cerebellum tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Myelin Basic Protein in human cerebellum tissue lysate.

Target description

~17 kDa observed; 18.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunshan He et al.
European journal of nuclear medicine and molecular imaging (2022-03-02)
Radical hysterectomy combined with pelvic lymphadenectomy is the standard treatment for early-stage cervical cancer, but unrecognized pelvic nerves are vulnerable to irreversible damage during surgery. This early clinical trial investigated the feasibility and safety of intraoperative near-infrared (NIR) fluorescence imaging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service