Skip to Content
Merck
All Photos(2)

Key Documents

ABN58

Sigma-Aldrich

Anti-phospho MAP1B (Thr1265) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Microtubule-associated protein 1B, MAP-1B, MAP1(X), MAP1.2, MAP1 light chain LC1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€484.00

€484.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€484.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (immunogen homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr1265)

Gene Information

mouse ... Map1B(17755)
rat ... Map1B(29456)

General description

Microtubule-associated protein 1B (MAP1B) is neuron-specific and is thought to play a part in controlling the neuronal cytoskeleton. MAP1B is made up of heavy and light chains and is involved in the microtubule and actin filament association. MAP1B may have an important role in axon formation by supporting microtubule dynamics and controlling cytoskeletal arrangement. MAP1B has also been associated with Netrin 1 signaling.

Specificity

Other homologies: Human (91% sequence homology).
This antibody recognizes MAP1B phosphorylated at Thr1265.

Immunogen

Epitope: Phosphorylated Thr1265
KLH-conjugated linear peptide corresponding to mouse MAP1B phosphorylated at Thr1265.

Application

Anti-phospho MAP1B (Thr1265) Antibody is an antibody against phospho MAP1B (Thr1265) for use in WB, IH & IC.
Immunohistochemistry Analysis: 1:600 dilution from a representative lot detected MAP1B in embryonic (E12) rat spinal cord tissue. (Image courtesy of Dr. Phillip R. Gordon-Weeks, King′s College London.)

Immunofluorescence Analysis: A representative lot was used by an independent laboratory in IF. (Trivedi, N., et al. (2005). Journal of Cell Science. 118:993-1005.)

Quality

Evaluated by Western Blot in S1 rat brain tissue lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected MAP1B on 10 µg of S1 rat brain tissue lysate.

Target description

~300 kDa observed

Analysis Note

Control
S1 rat brain tissue lysate

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service