Skip to Content
Merck
All Photos(1)

Key Documents

ABN484

Sigma-Aldrich

Anti-KCNK13 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Potassium channel subfamily K member 13, Tandem pore domain halothane-inhibited potassium channel 1, THIK-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€271.00

€271.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€271.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€271.00


Estimated to ship on26 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (immunogen homology)

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNK13(56659)

General description

Potassium channel subfamily K member 13 (KCNK13) is a member of the two pore domain potassium channel (TC 1.A.1.8) family. Members of this family are characterized by four transmembrane domains that may play an important role in helping to influence the resting membrane potential of cells. KCNK13 is expressed mainly in the brain with lower levels observed in kidneys.

Immunogen

KLH-conjugated linear peptide corresponding to human KCNK13.

Application

Detect Potassium channel subfamily K member 13 using this rabbit polyclonal antibody, Anti-KCNK13 Antibody validated for use in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay Analysis in rat brain tissue lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 0.5 µg/mL of this antibody detected KCNK13 in 15 µg of rat brain tissue lysate.

Target description

~62 kDa observed. Uncharacterized bands may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service