Skip to Content
Merck
All Photos(2)

Key Documents

ABN457

Sigma-Aldrich

Anti-Peptide YY Antibody

from chicken, purified by affinity chromatography

Synonym(s):

Peptide YY1, PYYPYY-I, Peptide tyrosine tyrosine, Peptide YY(3-36), PYY-II, Peptide YY

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€395.00

€395.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€395.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€395.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgY

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pyy(217212)
rat ... Pyy(287730)

General description

Peptide YY (UniProt P10082; also known as Peptide tyrosine tyrosine, PYY, PYY-I) is encoded by the PYY (also known as PYY1) gene (Gene ID 5697) in human. Peptide YY is initially produced as a preproprotein with a signal and a propeptide sequence (aa 1-28 and aa 68-97, respectively). There are two major forms of mature peptide YY corresponding to aa 29-64 and aa 31-64 of the prepro sequence, commonly known as YY(1-36) and YY(3-36), respectively. PYY is one of several well recognized circulating anorexigenic peptides, whose productions increase post-prandially. PYY and glucagon-like peptide 1 (GLP-1) are released from L cells that predominate in the distal ileum and colon. The melanocortin-4 receptor (MC4R) is reported to mediate PYY and GLP-1 release from enteroendocrine L cells upon melanocortin peptides stimulation.

Specificity

Reacts with prepro-, pro-, and mature peptide YY & peptide YY(3-36).

Immunogen

Epitope: Mature Peptide YY
KLH-conjugated linear peptide corresponding to the mature human peptide YY.

Application

Anti-Peptide YY Antibody, Cat. No. ABN457, is a highly specific chicken Polyclonal antibody, that targets Peptide YY and has been tested in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Peptide YY in rat colon tissue.

Quality

Evaluated by Western Blotting in rat small intestine tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Peptide YY in 10 µg of rat small intestine tissue lysate.

Target description

~11 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service