Skip to Content
Merck
All Photos(2)

Key Documents

ABN439

Sigma-Aldrich

Anti-PQBP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Polyglutamine-binding protein 1, PQBP-1, PQBP 1, 38 kDa nuclear protein containing a WW domain, Npw38, Polyglutamine tract-binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€322.00

€322.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€322.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PQBP1(10084)

General description

Polyglutamine-binding protein 1 (PQBP1), also known as PQBP-1, 38 kDa nuclear protein containing a WW domain, Npw38, Polyglutamine tract-binding protein 1, is encoded by the gene name PQBP1/ NPW38/ JM26. Recent studies have implied that PQBP1 may suppress the ability of POU3F2 to transactivate the DRD1 gene in a POU3F2 dependent manner. PQBP1 has also been shown to activate transcription directly, or via association with the transcription machinery. PQBP1 might be involved in ATXN1 mutant-induced cell death. PQBP1 has been shown to interact with POU3F2/Brn-2, ATXN1, TXNL4A, HTT and AR. Interaction with ATXN1 correlates positively with the length of the polyglutamine tract. In addition, PQBP1 interacts with RNA polymerase II large subunit in a phosphorylation-dependent manner. PQBP1 forms a ternary complex with ATXN1 mutant and phosphorylated RNA polymerase II. PQBP1 is widely expressed with high levels in heart, skeletal muscle, pancreas, spleen, thymus, prostate, ovary, small intestine and peripheral blood leukocytes. Mutations in PQBP1 have been associated with Renpenning syndrome 1 (RENS1), an X-linked mental retardation syndrome characterized by mental retardation, microcephaly, short stature, and small testes.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to the unknown of Human PQBP1.

Application

Anti-PQBP1 is an antibody against PQBP1 for use in WB, IH.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PQBP1 in human cardiac myocytes and pancreatic tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in Human Hippocampus tissue lysate.

Western Blotting Analysis: A 0.5 µg/mL dilution of this antibody detected PQBP1 in 10 µg of Human Hippocampus tissue lysate.

Target description

~38 kDa observed. Predicted to cross-react to Isoforms 1, 2, 3, and 6. Maybe detected upon longer exposure times.

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service