Skip to Content
Merck
All Photos(3)

Key Documents

ABN433

Sigma-Aldrich

Anti-Corticotropin-Releasing Factor Receptor 2 Antibody

1.0 mg/mL, from rabbit

Synonym(s):

Corticotropin-releasing factor receptor 2, CRF-R-2, CRF-R2, CRFR-2, Corticotropin-Releasing Factor Receptor 2, Corticotropin-releasing hormone receptor 2, CRH-R-2, CRH-R2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€435.00

€435.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), hamster (based on 100% sequence homology), primate (based on 100% sequence homology)

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CRHR2(1395)

General description

Corticotropin-Releasing Factor Receptor 2 (CRFR2) is a G-protein coupled receptor for CRH (Corticotropin-releasing factor), UCN (urocortin), UCN2 and UCN3. Ligand binding activates adenylate cyclase leading to an increase in cAMP levels and subsequent gene activation. CRFR2 binds Urocortin (UCN) with the highest affinity. CRFR2 is localized to the plasma membrane, and CRFR2 has numerous isoforms but their functional importance remains to be discovered. Latest research is focused upon antagonists of the receptors for CRF. Astressin2-B, which is specific for CRFR2 and Astressin B which binds both CRFR1 and CRFR2 are two recent peptide antagonists. The research hopes to develop treatments for many CRF-dependent diseases in the periphery such as irritable bowel syndrome and even perhaps various social behaviors such as anxiety and anti-social activities.

Immunogen

Epitope: Near N-terminal
KLH-conjugated linear peptide corresponding to human Corticotropin-Releasing Factor Receptor 2 near the N-terminal.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Corticotropin-Releasing Factor Receptor 2 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Corticotropin-Releasing Factor Receptor 2.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected Corticotropin-Releasing Factor Receptor 2 in 10 µg of human brain tissue lysate.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Corticotropin-Releasing Factor Receptor 2 in human colon and Bowman′s capsule epithelial cells.

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Corticotropin-Releasing Factor Receptor 2 in 10 µg of human colon tissue lysate.

Target description

~49 kDa observed. Uniprot describes 5 isoforms produced by alternative splicing from ~45-65 kDa

Linkage

Replaces: AB9420

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge A Rodriguez et al.
Cellular and molecular gastroenterology and hepatology, 1(6), 610-630 (2015-10-27)
Chronic inflammation promotes development and progression of colorectal cancer (CRC). We explored the distribution of Corticotropin-Releasing-Hormone (CRH)-family of receptors and ligands in CRC and their contribution in tumor growth and oncogenic EMT. mRNA expression of CRH-family members was analyzed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service