Skip to Content
Merck
All Photos(1)

Key Documents

ABN351

Sigma-Aldrich

Anti-LGI3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat LGI family member 3, LGI1-like protein 4, Leucine-rich glioma-inactivated protein 3, LGI3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGI3(203190)

General description

The leucine-rich, glioma inactivated gene 3 (LGI3) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI3 have not been found to be associated with ADTLE. LGI3 expression is induced in rat astrocyte cultures by the amyloid beta (Abeta) peptide and accumulated on neuronal plasma membranes of aged monkey brains and co-localized with Abeta.

Specificity

Two isoforms of LGI3 are known to exist; this LGI3 antibody will recognize both.

Immunogen

KLH-conjugated linear peptide corresponding to human LGI3.

Application

Detect the LGI3 protein using this Anti-LGI3 validated for use in WB.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected LGI3 in 15 µg of human brain tissue lysate.

Target description

~62 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service