Skip to Content
Merck
All Photos(2)

Key Documents

ABN292

Sigma-Aldrich

Anti-DDAH2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

N(G),N(G)-dimethylarginine dimethylaminohydrolase 2, DDAH-2, Dimethylarginine dimethylaminohydrolase 2, DDAHII, Dimethylargininase-2, Protein G6a, S-phase protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€263.00

€263.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€263.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€263.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDAH2(23564)

General description

Dimethylarginine dimethylaminohydrolase 2 (DDAH2) is also called N(G),N(G)-dimethylarginine dimethylaminohydrolase 2 (DDAH-2), DDAHII, Dimethylargininase-2, Protein G6a and S-phase protein. DDAH2 is involved in nitric oxide generation through the hydrolyzation of N(G),N(G)-dimethyl-L-arginine (ADMA) and N(G)-monomethyl-L-arginine (MMA) which are NOS inhibitors. DDAH2 is expressed in heart, placenta, lung, liver, skeletal muscle, kidney and pancreas.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human DDAH2.

Application

Detect Dimethylarginine dimethylaminohydrolase 2 using this rabbit polyclonal antibody, Anti-DDAH2 Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected DDAH2 in human kidney and human cardiac myocytes tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human heart tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected DDAH2 in 10 µg of human heart tissue lysate.

Target description

~36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service