Skip to Content
Merck

Skip To

ABN2300A4

Neuro-Chrom Pan Neuronal Marker Antibody-Rabbit, Alexa488 conjugate

Neuro-Chrom, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on13 October 2026fromMerck Life Science BV/SRL

€638.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
ALEXA FLUOR 488
Clone:
polyclonal
Application:
ICC, IF, IHC
Citations:
2

€638.00


Estimated to ship on13 October 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

ALEXA FLUOR 488

clone

polyclonal

species reactivity

mouse, rat

manufacturer/tradename

Neuro-Chrom

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibodies to neuronal proteins have become critical tools for identifying neurons and discerning morphological characteristics in culture and complex tissue. While the labeling from classic histological techniques such as Golgi staining and modern molecular approaches such as GFP constructs yield excellent cytoarchitectural detail, these approaches are technically challenging and impractical for many neuroscience research needs. Neuron-specific antibodies are convenient precision tools useful in revealing cytoarchitecture, but are limited to the protein target distribution within the neuron, which may differ greatly from nucleus to soma to dendrite and axon. To achieve as complete a morphological staining as possible across all parts of neurons, Millipore has developed a polyclonal antibody blend that reacts against key somatic, nuclear, dendritic, and axonal proteins distributed across the pan-neuronal architecture that can then be detected by a single secondary antibody. This antibody cocktail has been validated in diverse methods, cell culture and immuno-histochemistry, giving researchers a convenient and specific qualitative and quantitative tool for studying neuronal morphology.

Immunogen

Epitope: Whole Neuron Marker

Application

Neuro-Chrom Pan Neuronal Marker-Rabbit, Alexa488 conjugate is an antibody targeting the Pan Neuronal Marker protein, validated for use in ICC, IHC & IF.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Biochem/physiol Actions

Cat. # ABN2300A4 is specific to axons (neurites), dendrites, nucleus, and the cell body of neurons.
Reactivity with other species has not been determined.

Physical form

Polyclonal antibody cocktail blend conjugated to Alexa Fluor 488 in PBS buffer with 15mg/mL BSA and 0.05% NaN3

Preparation Note

Maintain at 2-8°C for up to 1 year from date of receipt.

Analysis Note

Control
Rat E18 cortex primary neuron cell culture or mouse adult brain cryosections.
Routinely tested Rat E18 cortex primary neurons in Immunocytochemistry.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN2300C3ABN2300NS340
description

Neuro-Chrom, from rabbit

description

Neuro-Chrom, from rabbit

description

Neuro-Chrom, from rabbit

description

Milli-Mark ChromaPan Neuronal Marker-ORC is an antibody targeting the ORC protein, validated for use in ICC & IF.

conjugate

ALEXA FLUOR 488

conjugate

CY3 conjugate

conjugate

-

conjugate

-

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

mouse, rat

species reactivity

mouse, rat

species reactivity

rat, mouse

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunofluorescence: suitable

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable

technique(s)

immunocytochemistry: suitable, immunofluorescence: suitable


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters