Skip to Content
Merck
All Photos(3)

Key Documents

ABN2261

Sigma-Aldrich

Anti-GAS2L1

from rabbit

Synonym(s):

GAS2-like protein 1, GAS2-related protein on chromosome 22, Growth arrest-specific protein 2-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€115.00
100 μG
€313.00

€115.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€115.00
100 μG
€313.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€115.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GAS2L1(10634)

General description

GAS2-like protein 1 (UniProt: Q99501-1; also known as GAS2-related protein on chromosome 22, Growth arrest-specific protein 2-like 1, GAS2L1) is encoded by the GAS2L1 (also known as GAR22) gene (Gene ID: 10634) in human. GAS2L1 belongs to the growth-arrest-specific 2 (GAS2) family, whose members mediate the crosstalk between filamentous actin (F-actin) and microtubules. Three members of this family, known as GAS2L1, GAS2L2, and GAS2L3, are shown to be differentially involved in mediating the crosstalk between F-actin and microtubules. GAS2L1 is expressed in testis and brain, GAS2L2 is exclusively expressed in skeletal muscle, and GAS2L3 is found in many different cell types. Although they all localize to actin and microtubules, only the exogenous expression of GAD2L1 and GAS2L2 affect microtubule stability, dynamics, and guidance along actin stress fibers. GAS2L1 is reported to associate with the proximal end of mature centrioles and participates in centriole dynamics and centrosome disjunction. Loss of GAS2L1 can inhibit centrosome disjunction in the G2 phase. On the other hand, overexpression of GAS2L1 can induce premature centrosome separation, which also requires its association with actin, microtubules, and the microtubule end-binding proteins. (Ref.: Stroud MJ et al. (2014). J. Cell Sci. 127(12); 2672-2682; AU, FKC et al. (2017). Dev. Cell 40(1); 81-94).

Specificity

This rabbit polyclonal antibody GAS2-like protein 1 (GAS2L1) in human cells. It targets an epitope with in 188 amino acids from the C-terminal half.

Immunogen

Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 188 amino acids from the C-terminal region of human GAS2-Like protein 1.

Application

Anti-GAS2L1, Cat. No. ABN2261, is a highly specific rabbit polyclonal antibody that targets GAS2-like protein 1 (Gas2L1) and has been tested for use in Immunofluorescence, Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected GAS2L1 in Western Blotting applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).

Immunoprecipitation Analysis: 1 µg from a representative lot detected GAS2L1 in U2OS cell lysate (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology).

Immunofluorescence Analysis: A representative lot detected GAS2L1 in Immunofluorescence applications (Au, F.K., et. al. (2017). Dev Cell. 40(1):81-94).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected GAS2L1 in lysate from HeLa cells, but not in lysate from GAS2L1 knockout cells (Courtesy of Dr Robert Z. Qi and Dr. Franco K.C. Au at Hong Kong University of Science and Technology).

Quality

Evaluated by Western Blotting in HeLa and GAS2L1 knockout lysate.

Western Blotting Analysis: 0.9 µg/mL of this antibody detected GAS2-like protein 1 (Gas2L1) in HeLa cell lysate. It did not detect GAS2L1 in lysate from knockout cells.

Target description

~78 kDa observed; 72.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 10% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service