Skip to Content
Merck
All Photos(1)

Key Documents

ABN2246

Sigma-Aldrich

Anti-PTP zeta/Phosphacan

from rabbit

Synonym(s):

Receptor-type tyrosine-protein phosphatase zeta, EC:3.1.3.48, R-PTP-zeta, 3F8 chondroitin sulfate proteoglycan, 3H1 keratan sulfate proteoglycan, Phosphacan

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
100 μL
€330.00

€130.00


Estimated to ship on18 February 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
100 μL
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on18 February 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Receptor-type tyrosine-protein phosphatase zeta (UniProt: Q62656; also known as EC:3.1.3.48, R-PTP-zeta, 3F8 chondroitin sulfate proteoglycan, 3H1 keratan sulfate proteoglycan, Phosphacan) is encoded by the Ptprz1 (also known as Ptprz, Ptpz) gene (GeneID: 25613) in rat. It is a soluble nervous tissue-specific proteoglycan that is thought to regulate axonal outgrowth. It is synthesized by glia and binds to neurons and to the neural cell adhesion molecules tenascin, N-CAM or NG-CAM but not to laminin and fibronectin. Phosphacan acts as a potent inhibitor of cell adhesion and neurite outgrowth and negatively regulates oligodendrocyte precursor proliferation in the embryonic spinal cord. It is required for normal differentiation of the precursor cells into mature, fully myelinating oligodendrocytes. RPTPzeta has an extracellular domain (aa 25-1637), a transmembrane domain (aa 1638-1663) and a cytoplasmic domain (aa 1664-2316). It is produced with a signal peptide (aa 1-24) that is cleaved to generate the mature form.

Specificity

This rabbit polyclonal antibody specifically detects PTP zeta/Phosphacan in rat brain tissue.

Immunogen

Epitope: unknown
Full length (aa 1-2316) purified Receptor-type tyrosine-protein phosphatase zeta (Phosphacan) isolated from 7-day fetal rat brain.

Application

Anti-phosphacan, Cat. No. ABN2246 is a rabbit polyclonal antibody that has been tested for use in Immunohistochemistry (Paraffin) for the detection of Receptor-type tyrosine-protein phosphatase zeta.
Immunohistochemistry Analysis: A representative lot detected PTP zeta/Phosphacan in E16 rat spinal cord (Milev, P., et. al. (1994). J Cell Biol. 127(6 Pt 1):1703-15), rat brain (Haunso, A., et. al. (1999). Brain Res. 834(1-2):219-22), and 21-day postnatal rat retina, 2-week postnatal rat cerebellum, E13 rat spinal cord (Milev, P., et. al. (1998). J Biol Chem. 273(12):6998-7005).
Research Category
Cell Structure

Quality

Evaluated by Immunohistochemistry (Paraffin) in rat cerebral cortex and rat hippocampus tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected PTP zeta/Phosphacan in rat cerebral cortex and rat hippocampus tissue.

Target description

255.34 kDa calculated.

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.02% sodium azide.
Rabbit polyclonal antiserum with 0.02% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service