Skip to Content
Merck
All Photos(2)

Key Documents

ABN213

Sigma-Aldrich

Anti-IL-12 alpha Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-12 subunit alpha, IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35, NK cell stimulatory factor chain 1, NKSF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€435.00

€435.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL12A(3592)

General description

Interleukin-12 alpha (IL-12A) is also known as Cytotoxic lymphocyte maturation factor 35 kDa subunit (CLMF p35), IL-12 subunit p35, or NK cell stimulatory factor chain 1 (NKSF-1). IL-12A is a cytokine that, along with IL12B, forms a disulfide-linked heterodimer interleukin IL-12. IL-12 alpha functions as a regulator of T cell mediated cytotoxicity, interferon-gamma production, lymphocyte proliferation and activation of natural killer cells.

Specificity

This antibody recognizes the near C-terminus of IL-12 alpha

Immunogen

KLH-conjugated linear peptide corresponding to the near C-terminus of human IL-12 alpha.

Application

Immunohistochemistry Analysis: A 1:16,000-32,000 dilution from a representative lot detected IL-12 alpha in human tonsil and human brain tissue.
This IL-12 alpha antibody is validated for use in WB & IHC for the detection of the IL-12 alpha protein.

Quality

Evaluated by Western Blotting in human lymph node tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected IL-12 alpha in 10 µg of human lymph node tissue lysate.

Target description

~25 kDa observed

Linkage

Replaces: MABN1084

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service