Skip to Content
Merck
All Photos(2)

Key Documents

ABN204

Sigma-Aldrich

Anti-MOB kinase activator 3B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

MOB kinase activator 3B, Mob1 homolog 2b, Mps one binder kinase activator-like 2B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€279.00

€279.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€279.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€279.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MOB3B(79817)

General description

MOB kinase activator 3B (MOB3B) is also known as Mob1 homolog 2b or Mps one binder kinase activator-like 2B (MOBKL2B). MOB kinase activator 3B is thought to regulate the activity of kinases based on similarity with the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase critical to spindle pole body duplication and mitotic checkpoint regulation.

Specificity

Other homologies: Zebrafish and Xenopus (85% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human MOB kinase activator 3B.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected MOB kinase activator 3B in mouse midbrain and mouse hippocampus tissue.
This MOB kinase activator 3B antibody is validated for use in WB & IHC for the detection of the MOB kinase activator 3B protein.

Quality

Evaluated by Western Blotting in MOB kinase activator 3B transfected cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected MOB kinase activator 3B in 10 µg of MOB kinase activator 3B transfected cell lysate.

Target description

~25 kDa observed

Analysis Note

Control
MOB kinase activator 3B transfected cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service