Skip to Content
Merck
All Photos(1)

Key Documents

ABN1640

Sigma-Aldrich

Anti-Prodynorphin/Leumorphin (dynorphin B29)

serum, from rabbit

Synonym(s):

Preproenkephalin-B, Proenkephalin-B, Beta-neoendorphin-dynorphin, Leumorphin, Dynorphin B-29

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€373.00

€373.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 μL
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PDYN(5173)
rat ... Pdyn(29190)

General description

Proenkephalin-B (UniProt P06300; also known as Beta-neoendorphin-dynorphin, Preprodynorphin) is encoded by the Pdyn gene (Gene ID 29190) in rat species. Proenkephalin B is an opioid polypeptide hormone involved with chemical signal transduction and cell communication. Prodynorphin is a basic building-block of endorphins, the chemical messengers in the brain that appear most heavily involved in the anticipation and experience of pain and the formation of deep emotional bonds critical in learning and memory. Prodynorphin is produced with a signal peptide (a.a. 1-21) and a propeptide (a.a. 22-163) sequence. Following the removal of the signal and propeptide sequences, the protein is further processed by proeolytic cleavages to form Alpha-neoendorphin (a.a. 166-175), Beta-neoendorphin (a.a. 166-174), Big dynorphin (a.a. 202-233; Big Dyn), Dynorphin A(1-17) (a.a. 202-218; Dyn-A17; Dynorphin A); Dynorphin A(1-13) (a.a. 202-214), Dynorphin A(1-8) (a.a. 202-209), Leu-enkephalin (a.a. 202-206 & 221-225), Leumorphin (Dynorphin B-29; a.a. 221-248), and Rimorphin (a.a. 221-233; Dynorphin B; Dyn-B; Dynorphin B(1-13)).

Specificity

This polyclonal antiserum recognizes preproenkephalin-B (a.a. 1-248 of rat sequence), proenkephalin-B (a.a. 22-248 of rat sequence), and leumorphin/dynorphin B-29 (a.a. 221-248 of rat sequence), but not other peptides derived from Proenkephalin-B.

Immunogen

Epitope: Leumorphin peptide.
Linear peptide corresponding to rat leumorphin (dynorphin B-29) C-terminal half sequence.

Application

Anti-Prodynorphin Antibody is an antibody against Leumorphin (dynorphin B29) for use in Western Blotting.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected leumorphin (dynorphin B29) in 10 µg of human brain tissue lysate.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected prodynorphin in 10 µg of rat brain tissue lysate.

Target description

~36/3.5 kDa oberved. 28.08/28.06/28.39 kDa (Rat/Mouse/Human preproenkephalin-B), 25.85/25.76/26.28 kDa (Rat/Mouse/Human proenkephalin-B), 3.320/3.320/3.352 kDa (Rat/Mouse/Human leumorphin) calculated. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service