Skip to Content
Merck
All Photos(2)

Key Documents

ABN1455

Sigma-Aldrich

Anti-NKX6.2

from rabbit, purified by affinity chromatography

Synonym(s):

Homeobox protein NK-6 homolog B, Homeobox protein Nkx-6.2, NKX6.2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NKX6-2(84504)

General description

Homeobox protein Nkx-6.2 (UniProt Q9C056; also known as Homeobox protein NK-6 homolog B) is encoded by the NKX6-2 (also known as GTX, NKX6B) gene (Gene ID 84504) in human. Mammalian NKX gene products and their orthologs in other species are known to regulators of neuronal cell fate. Both the ventral nervous system defective (vnd) gene product in Drosophila and murine NKX proteins in spinal cord direct progenitors to adopt specific ventral neural identities. In mice, NKX6.2 is expressed in a small subset of ventral neural progenitors in a restricted spatial pattern at the border between the medial and caudal ganglionic eminence (MGE and CGE). NKX6,2 is required for specification of V1 interneurons in the spinal cord and progenitors are found to adopt a more dorsal V0 identity in the absence of NKX6,2. In addition, NKX6.2-positive brain region is shown to give rise to both parvalbumin (PV) and somatostatin (SST) cortical interneurons and in particular be enriched in a subpopulation of SST cortical interneurons that coexpress calretinin (CR).

Specificity

This rabbit polyclonal antibody detected the exogenously expressed human NKX6.2 in transfected HEK293T cells, while no immunoreactive bands were detected in lysates from untransfected cells. The immunogen sequence lies within the region C-terminal to the Homeobox and is 100% conserved among human, mouse, and rat species.

Immunogen

KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human/mouse/rat NKX6.2.

Application

Anti-NKX6.2, Cat. No. ABN1455, is a highly specific rabbit polyclonal antibody that targets Homeobox protein Nkx-6.2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected NKX6.2 in human cerebral cortex and kidney tissue sections.

Western Blotting Analysis: 1 µg/mL from a representative lot detected Myc-DDK-tagged human NKX6.2 in 10 µg of lysate from transfected HEK293T cells.
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in human pancreas tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected NKX6.2 in human pancreas tissue section.

Target description

~29 kDa observed. 29.26 kDa (human) and 29.36 kDa (mouse and rat) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service