Skip to Content
Merck
All Photos(1)

Key Documents

ABN117

Sigma-Aldrich

Anti-PDE4D3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

cAMP-specific 3′,5′-cyclic phosphodiesterase 4D, DPDE3, PDE43

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, rhesus macaque

species reactivity (predicted by homology)

human (immunogen homology), rhesus monkey (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDE4D(5144)

General description

Phosphodiesterase 4D3 (PDE4D3) is a member of the PDE cyclic nucleotides. PDE4D3 is known to be regulated by protein kinase C (PKC)-Raf-MEK-ERK and also by cAMP in vascular smooth muscle cells. PDE4D3 is a direct target of the MSH/cAMP/MITF pathway and may play a role in melanocyte differentiation. PDE4D3 is essential to have equilibrium in relaxing and contracting cues to airway smooth muscle. Down-regulation of PDE4D3 may be associated with analgesic and anti-inflammatory effects. PDE4D3 has also been linked to the risk of stroke, and in particular to polygenic stroke.

Specificity

This antibody recognizes PDE4D3 at the N-terminus.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human PDE4D3.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-PDE4D3 Antibody is validated for use in WB for the detection of PDE4D3.

Quality

Evaluated by Western Blot in mouse brain membrane tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected PDE4D3 on 10 µg of mouse brain membrane tissue lysate.

Target description

~95 kDa observed. UniProt describes 12 isoforms produced by alternative splicing.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain membrane tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinghua Cheng et al.
Diabetes, 62(12), 4088-4097 (2013-08-27)
In utero exposure to gestational diabetes mellitus (GDM) is associated with an increased risk of type 2 diabetes and cardiovascular disease in later life, yet the underlying mechanisms remain to be elucidated. We examined the effects of GDM on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service