Skip to Content
Merck
All Photos(2)

Key Documents

ABN105

Sigma-Aldrich

Anti-Sox11 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Transcription factor SOX-11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€484.00

€484.00


Estimated to ship on05 May 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1337

Request a Bulk Order

Select a Size

Change View
100 μG
€484.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€484.00


Estimated to ship on05 May 2025Details

A recombinant, preservative-free antibody is available for your target. Try ZRB1337

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (tissue)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOX11(6664)

General description

SRY box-containing 11 (Sox11) is a neural transcription factor belonging to the DNA-binding high-mobility group (HMG) box protein super family. It is highly expressed during embryonic development and regulates genes involved in neurite growth and survival. Sox11 plays an important role in the formation of sensory neurons and organogenesis. Sox11-deficient mice show defects in multiple organs including the lung, heart, and pancreas. Sox11 also plays a critical role as a tumor suppressor. Epigenetic suppression of the Sox11 gene, primarily through promoter hypermethylation, contributes to various forms of neoplasms. In adults, Sox11 may be involved in the regeneration of neurons after nerve injury by inducing the expression of the small proline rich protein 1a gene (Sprr1a).

Immunogen

KLH-conjugated linear peptide corresponding to human Sox-11.

Application

Anti-Sox11 Antibody detects level of Sox11 & has been published & validated for use in 0.
Immunohistochemistry: A 1:1,000 dilution from a representative lot detected Sox-11 in normal mouse hippocampus and cerebellum tissues.

Western Blot Analysis: A representative lot detecte Sox-11 in human brain tissue lysate.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 0.5 mg/mL of this antibody detected Sox-11 in 10 mg of mouse brain tissue lysate.

Target description

~57 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyan Cheng et al.
Nature communications, 13(1), 4418-4418 (2022-07-30)
The inability of neurons to regenerate long axons within the CNS is a major impediment to improving outcome after spinal cord injury, stroke, and other CNS insults. Recent advances have uncovered an intrinsic program that involves coordinate regulation by multiple
Elena Spina et al.
Nature communications, 13(1), 1421-1421 (2022-03-19)
Gpr125 is an orphan G-protein coupled receptor, with homology to cell adhesion and axonal guidance factors, that is implicated in planar polarity and control of cell movements. By lineage tracing we demonstrate that Gpr125 is a highly specific marker of
Rotem Ben-Tov Perry et al.
Cell reports, 42(10), 113168-113168 (2023-09-24)
Long noncoding RNAs (lncRNAs) are expressed in many brain circuits and types of neurons; nevertheless, their functional significance for normal brain functions remains elusive. Here, we study the functions in the central nervous system of Silc1, an lncRNA we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service