Skip to Content
Merck
All Photos(2)

Key Documents

ABN1024

Sigma-Aldrich

Anti-BMP-9 (GDF-2)

from rabbit, purified by affinity chromatography

Synonym(s):

Bone morphogenetic protein 9, Growth/differentiation factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€317.00

€317.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
200 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


In StockDetails


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GDF2(2658)

General description

Growth/differentiation factor 2 (UniPort: Q9UK05; also known as GDF-2, Bone morphogenetic protein 9, BMP-9) is encoded by the GDF2 (also known as BMP9) gene (Gene ID: 2658) in human. BMP-9 is a member of the TGF-beta family that forms a signaling complex with two type 1 and two type II receptors. It is synthesized with a signal peptide of (aa 1-22) and a propeptide (aa 22-319). The signal peptide is cleaved in the mature protein. The propeptide of BMP-9 does not interfere with the biological activity of BMP-9 and is believed to remain associated with the mature peptide. BMP9 is constitutively secreted from the liver in an active form into the circulation. BMP-9 signals through high affinity activing receptor-like kinase 1 (ALK1) in endothelial cells and maintains endothelial homeostasis. It is also considered as a potent osteogenic signaling factor in mesenchymal stem cells. BMP-9 is also known to induce an increase in collagen, fibronectin, and connective tissue growth factor expression associated with Smad 1/5/8, Smad 2/3, and Erk1/2 activation. It is also reported to promote the acquisition of the cholinergic phenotype in basal forebrain cholinergic neurons during development and protects these neurons. Defects in BMP9 gene are known to cause Telangiectasia, hereditary hemorrhagic disease that leads to vascular dysplasia leading to dilation of permanent blood vessels and arteriovenous malformations of skin, mucosa, and viscera and causes recurrent epistaxis and gastro-intestinal hemorrhage (Ref.: Wei, Z et al. (2014). J. Biol. Chem. 289 (45); 31150 31159; Munoz-Felix, JM et al. (2016). Cell Signal 28(9): 1252-1261).

Specificity

This rabbit polyclonal antibody detects human Bone morphogenetic protein 9 (BMP-9). It targets an epitope within 110 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 110 amino acids from the C-terminal region of human Bone morphogenetic protein 9 (BMP-9).

Application

Anti-BMP-9 (GDF-2), Cat. No. ABN1024, is a highly specific rabbit polyclonal antibody that targets Bone morphogenetic protein 9 (Growth/differentiation factor 2) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BMP-9 (GDF-2) in human liver and human liver cancer tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected BMP-9 (GDF-2) in 10 µg of human liver tissue lysate.

Target description

~45 kDa observed; 47.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine, 0.15 M NaCl, pH7.4 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service