ABN1016
Anti-CCM3/PDCD10
from rabbit, purified by affinity chromatography
Synonym(s):
Programmed cell death protein 10, Cerebral cavernous malformations 3 protein, TF-1 cell apoptosis-related protein 15
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
Xenopus (based on high sequence homology), rat (based on high sequence homology), mouse (based on high sequence homology)
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... PDCD10(11235)
General description
Specificity
Immunogen
Application
Neuroscience
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CCM3/PDCD10 in 100 µg of lysate from untreated, but not CCM3 shRNA-treated, EA.hy926 human umbilical vein endothelial cells (Courtesy of Kyle M. Draheim, Ph.D., Xiaofeng Li, Ph.D., Rong Zhang, David A.Calderwood, Ph.D. and Titus J. Boggon, Ph.D., Yale University, U.S.A.).
Quality
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected CCM3/PDCD10 in 10 µg of EA.hy926 human umbilical vein endothelial cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service