Skip to Content
Merck
All Photos(1)

Key Documents

ABF998

Sigma-Aldrich

Anti-IspH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 34.78 kDa
observed mol wt ~35 kDa

purified by

using Protein A

species reactivity

E. coli

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

bacterial ... ispH(944777)

General description

4-hydroxy-3-methylbut-2-enyl diphosphate reductase (UniProt: P62623; also known as EC:1.17.7.4, HMBPP reductase, 1-hydroxy-2-methyl-2-(E)-butenyl 4-diphosphate reductase) is encoded by the ispH (also known as lytB, yaaE, b0029, JW0027) gene (Gene ID: 944777) in E. coli. IspH is an enzyme that catalyzes the conversion of 1-hydroxy-2-methyl-2-(E)-butenyl 4-diphosphate (HMBPP) into a mixture of isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP). It contains a central iron-sulfur cluster that serves as an electron transfer cofactor in the active site. IspH acts in the terminal step of the methyl erythritol phosphate (MEP) pathway for isoprenoid precursor biosynthesis that is essential for survival of Gram-negative bacteria, mycobacteria, and apicomplexans. It operates at an optimum pH of 7.0. Its substrate binding site is localized in amino acids 225-227. Since MEP pathway is absent in human, inhibitors of IspH are attractive targets for the development of antibiotics. More recently, ester-based prodrug of C23 analog have been described that inhibit IspH and are highly potent against Gram-negative bacteria. (Ref.: Singh, KS., et al. (2021). Nature 589(7843); 597-602; Grawert, T., et al. (2004). J. Am. Chem. Soc. 126(40); 12847-12855).

Specificity

This rabbit polyclonal antibody specifically detects 4-hydroxy-3-methylbut-2-enyl diphosphate reductase (IspH) in E.coli.

Immunogen

Purified E. coli IspH protein was mixed with the KLH conjugate and Freud s complete adjuvant.

Application

Quality Control Testing

Evaluated by Western Blotting in E. coli lysates of strain CGSC8074 (positive) with conditional knockdown of IspH.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected IspH in lysate from E. coli, strain CGSC8074, but not in lysate from E.coli with conditional knockdown of IspH.

Tested Applications

Western Blotting Analysis: A representative lot detected IspH in Western Blotting applications (Singh, K., et al. (2021). Nature. 589(7843):597-602).

Enzyme Immunoassay Analysis (ELISA): A representative lot detected IspH in ELISA applications (Singh, K., et al. (2021). Nature. 589(7843):597-602).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-IspH, Cat. No. ABF998, is a rabbit polyclonal antibody that detects 4-hydroxy-3-methylbut-2-enyl diphosphate reductase (IspH) and is tested for use in ELISA and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service