Skip to Content
Merck
All Photos(2)

Key Documents

ABF212

Sigma-Aldrich

Anti-CIITA Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

MHC class II transactivator, MHC2TA, C2TA, NLRA, CIITAIV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€341.00

€341.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€341.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CIITA(4261)

General description

CIITA, also known as MHC class II transactivator (MHC2TA), is located in the nucleus and is an essential factor for transcriptional activity of the HLA class II promoter. CIITA is suggested to be a positive regulator of class II major histocompatibility complex gene transcription and is referred to as the "master control factor" for the expression of these genes. Additionally, CIITA contains an acidic transcriptional activation domain, four LRRs (leucine-rich repeats) and a GTP binding domain. CIITA binds with GTP and uses GTP binding to facilitate its own transport into the nucleus. Once in the nucleus it does not bind with DNA but rather uses an intrinsic acetyltransferase (AT) activity to act in a coactivator-like fashion. Furthermore, it may activate HLA class II transcription by modifying proteins that bind to the MHC class II promoter. CIITA has been shown to interact with ZXDA and ZXDC. Mutations in this gene have been associated with bare lymphocyte syndrome type II (also known as hereditary MHC class II deficiency or HLA class II-deficient combined immunodeficiency), increased susceptibility to rheumatoid arthritis, multiple sclerosis, and possibly myocardial infarction.

Immunogen

KLH-conjugated linear peptide corresponding to human CIITA.

Application

Detect MHC class II transactivator using this rabbit polyclonal antibody, Anti-CIITA Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected CIITA in human brain tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected CIITA in 15 µg of mouse brain tissue lysate.

Target description

~122 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service