Skip to Content
Merck
All Photos(2)

Key Documents

ABF184

Sigma-Aldrich

Anti-IL-21 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-21, IL-21, Za11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€356.00

€356.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€356.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€356.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL21(59067)

General description

Interleukin-21 (UniProt Q9HBE4; also known as IL-21, Za11) is encoded by the IL21 (also known as CVID11) gene (Gene ID 59067) in human. IL-21 is a type I four-alpha-helical bundle cytokine primarily produced by CD4+ T cells and NKT cells. The heterodimeric receptor IL-21R is dominantly expressed by B cells and shares the common cytokine receptor gamma chain (γc) with receptors for IL-2, IL-4, IL-7, IL-9, and IL-15. Studies indicate that IL-21 potently regulates both adaptive and innate immune responses and possesses widely pleiotropic functions. IL-21 has also been linked to autoimmune disorders, allergies, inflammatory diseases, as well as the pathogenesis of cancer. Human IL-21 is initially translated with an N-terminal 22 aa signal peptide sequence, which is later removed to produce the 133 aa mature IL-21.

Specificity

Expected to react with pro- and mature IL-21 of both isoform 1 and isoform 2 (IL-21iso).

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human IL-21 sequence near the C-terminus.

Application

Flow Cytometry Analysis: 3.0 µg of this antibody detected IL-21 in human peripheral blood mononuclear cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This rabbit Polyclonal Anti-IL-21 Antibody, Cat. No. ABF184 is validated for use in Western Blotting, Flow Cytometry, for the detection of IL-21.

Quality

Evaluated by Western Blotting in human IL-21 transfected HEK293 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected IL-21 in 10 µg of human IL-21 transfected HEK293 cell lysate.

Target description

~17 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biljana Bufan et al.
Life sciences, 301, 120617-120617 (2022-05-10)
Given that deprivation of noradrenaline acting on lymphocytes through β-adrenoceptor influences antibody response, the effects of propranolol treatment beginning two days before immunization with quadrivalent inactivated influenza vaccine (QIV) on IgG response and underlying cellular molecular mechanism in mice were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service