Skip to Content
Merck
All Photos(1)

Key Documents

ABF128

Sigma-Aldrich

Anti-Sharpin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Sharpin, Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, hSIPL1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€330.00

€330.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SHARPIN(81858)

General description

Sharpin, also known as Sharpin Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, hSIPL1, and encoded by the gene name SHARPIN/ SIPL1/ PSEC0216, is component of the LUBAC complex which conjugates linear polyubiquitin chains in a head-to-tail manner to substrates and plays a key role in NF-kappa-B activation and regulation of inflammation. LUBAC conjugates linear polyubiquitin to IKBKG and RIPK1 and is involved in activation of the canonical NF-kappa-B and the JNK signaling pathways. Linear ubiquitination mediated by the LUBAC complex interferes with TNF-induced cell death and thereby prevents inflammation. LUBAC is proposed to be recruited to the TNF-R1 signaling complex (TNF-RSC) following polyubiquitination of TNF-RSC components by BIRC2 and/or BIRC3 and to conjugate linear polyubiquitin to IKBKG and possibly other components contributing to the stability of the complex. Together with FAM105B/otulin, the LUBAC complex regulates the canonical Wnt signaling during angiogenesis. Sharpin is highly expressed in skeletal muscle and placenta and at lower levels in brain, heart, colon without mucosa, thymus, spleen, kidney, liver, small intestine, lung and peripheral blood leukocytes. Sharpin is up-regulated in various tumor tissues such as kidney, liver, ovary and pancreas tumors

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to the N-terminus of human Sharpin.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-Sharpin antibody is validated for use in WB, IP for the detection of Sharpin.
Western Blotting Analysis: A representative lot detected Sharpin in Splenic B cells were treated with an anti-CD40 antibody, anti-NEMO immunoprecipitates (IP) from TNF-a-treated MEFs HEK293T, Jurkat, cpdm MEFs, Cpdm MEFs transfected with control or HOIL-1L-specific siRNA, Brain lysates of Wt and cpdm mice, transfected cpdm MEFs, Lysates from Wt mice organsand splenic B and T cells from Wt and cpdm mice (Tokunaga, F., et al. (2010). Nature. 471:633-636).
Western Blotting Analysis: A representative lot detected Sharpin in HeLa cell lysate (Li, S., et al. (2013). Nature. 242-246).
Western Blotting Analysis: A representative lot detected Sharpin in HOIP-null MEFs, HOIP WT MEFs or HOIP N84A/Y93A MEFs (Takiuchi, T., et al. (2014). Genes to Cells. 1-19).
Western Blotting Analysis: A representative lot detected Sharpin in MEFs, BMDMs, and primary keratinocytes
from WT mice (Tamiya, H., et al. (2014). J. Immunology. 192:000-000).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Sharpin in HEK293T (Tokunaga, F., et al. (2010). Nature. 471:633-636).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Sharpin in 10 µg of HepG2 cell lysate.

Target description

~40 kDa observed. Anti-SHARPIN antibody demonstrated an approximately 40 kDa protein, the band is consistent with the predicted 39.9 kDa protein sequence for mouse. Uncharacterized band(s) may appear in some lysates.

Physical form

Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hironari Tamiya et al.
Cancers, 15(13) (2023-07-14)
Sarcoma is a rare type of cancer for which new therapeutic agents are required. Ferroptosis is a nonapoptotic cell death triggered by iron-mediated lipid peroxidation. We found that TFRC, an iron uptake protein, was expressed at higher levels in sarcoma
Xiao-Lin Shen et al.
The Journal of cell biology, 221(1) (2021-11-24)
Primary cilia transduce diverse signals in embryonic development and adult tissues. Defective ciliogenesis results in a series of human disorders collectively known as ciliopathies. The CP110-CEP97 complex removal from the mother centriole is an early critical step for ciliogenesis, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service