Skip to Content
Merck
All Photos(1)

Key Documents

ABF1070

Sigma-Aldrich

Anti-DRBP76/ILF3 Antibody

serum, from rabbit

Synonym(s):

Interleukin enhancer-binding factor 3, Double-stranded RNA-binding protein 76, DRBP76, M-phase phosphoprotein 4, MPP4, Nuclear factor associated with dsRNA, NFAR, Nuclear factor of activated T-cells 90 kDa, NF-AT-90, Translational control protein 80, TCP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€322.00

€322.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€322.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ILF3(3609)

General description

Interleukin enhancer-binding factor 3 (UniProt Q12906; also known as Double-stranded RNA-binding protein 76, DRBP76, dsRNA binding protein NFAR-2/MPP4, Interleukin enhancer binding factor 3 90kD, M-phase phosphoprotein 4, MPP4, NF-AT-90, NFAR, Nuclear factor associated with dsRNA, Nuclear factor of activated T-cells 90 kDa, TCP80, Translational control protein 80) is encoded by the ILF3 (also known as DRBF, NF90, MPHOSPH4) gene (Gene ID 3609) in human. DRBP76 is a dsRNA binding protein (DRBP) family member capable of binding both dsRNAs and highly structured single-stranded RNA molecules through its C-terminal region. DRBP76 influences the nucleus-cytoplasm export, stability, and translation of target mRNAs via direct interaction with their 3′ UTRs. DRBP76 is also reported to play both positive and negative regulatory roles in viral replications. DRBP76 is shown to interact with the 3′-terminal stem loop (SL) structure of the single-stranded RNA genome of the dengue virus and DRBP76 knockdown decreases production of functional dengue virus in cultured cells. On the other hand, DRBP76 can interact with full-length Zaire Ebola virus (EBOV) protein VP35 in the absence of exogenous dsRNA. Viral VP35 expression redistributes DRBP76 from the nucleus to the cytoplasm in the host cells and over-expression of DRBP76 impairs the function of the EBOV transcription/replication complex.

Specificity

Expected to react with all 7 spliced isoforms.

Immunogen

His-tagged recombinant protein corresponding to human DRBP76/ILF3.

Application

This Anti-DRBP76/ILF3 Antibody is validated for use in Western Blotting for the detection of DRBP76/ILF3.
Western Blotting Analysis: A representative lot detected DRBP76/ILF3 in the nuclear fractions prepared from the squamous cell carcinoma of the head and neck (SCCHN) cell lines PCI-15A and PCI-15B (Umemura, N., et al, (2012). Cancer Res. 72(1):45-55).
Western Blotting Analysis: A representative lot detected DRBP76/ILF3 in the nuclear fractions prepared from Wt11 (HEK293 expressing Flag-tagged wild-type TLR3) cells (Zhu, J., et al, (2010). J Immunol. 184(10):5768-5776; Sarkar, S.N., et al, (2004). Nat Struct Mol Biol. 11(11):1060-1067).

Quality

Evaluated by Western Blotting in MDA-MB-435 cell lysate.

Western Blotting Analysis: A 1:500 of this antibody detected DRBP76/ILF3 in 10 µg of MDA-MB-435 cell lysate.

Target description

~100 kDa A lower size band ~75 kDa may be seen in some lysates, most likely representing a smaller size variant or a non-specific band. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Martindale et al.
Bio-protocol, 10(2), e3488-e3488 (2020-01-20)
RNAs and RNA-binding proteins (RBPs) can interact dynamically in ribonucleoprotein (RNP) complexes that play important roles in controlling gene expression programs. One of the powerful ways to investigate changes in the association of RNAs with an RBP of interest is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service