Skip to Content
Merck
All Photos(2)

Key Documents

ABE95

Sigma-Aldrich

Anti-PRDM12 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

PR domain zinc finger protein 12, PR domain-containing protein 12, arg methyltransferase, arginine methyltransferase, PR domain family

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€498.00

€498.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€498.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€498.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRDM12(59335)

General description

Histone methylation occurs on arginine and lysine residues and is catalyzed by two families of proteins, the protein arginine methyltransferase family and the SET-domain-containing methyltransferase family. Five members have been identified in the arginine methyltransferase family. About 27 are grouped into the SET-domain family, and another 17 make up the PR domain family that is related to the SET domain family. The retinoblastoma protein-interacting zinc finger gene RIZ1 is a tumor suppressor gene and the founding member of the PR domain family. PRDM12 (PR domain zinc finger protein 12) also acts as a tumor suppressor and is linked to the pathogenesis of chronic myeloid leukemia. PRDM12 is also thought to play a role in transcriptional regulation.

Immunogen

KLH-conjugated linear peptide corresponding to human PRDM12.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PRDM12 in NIH/3T3 and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-PRDM12 Antibody (Rabbit Polyclonal Antibody) validated in WB, ICC to detect PRDM12 also known as PR domain zinc finger protein 12, arginine methyltransferase.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 3 μg/mL of this antibody detected PRDM12 on 10 µg of human liver tissue lysate.

Target description

~37 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clara E Hael et al.
Molecular metabolism, 34, 43-53 (2020-03-18)
Regulation of food intake and energy balance depends on a group of hypothalamic neurons that release anorexigenic melanocortins encoded by the Pomc gene. Although the physiological importance of central melanocortins is well appreciated, the genetic program that defines the functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service