Skip to Content
Merck
All Photos(3)

Key Documents

ABE602

Sigma-Aldrich

Anti-SATB1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DNA-binding protein SATB1, Special AT-rich sequence-binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€322.00

€322.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€322.00


Estimated to ship on04 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SATB1(6304)

General description

Special AT-rich sequence-binding protein 1 (SATB1) contains 2 CUT DNA-binding domains and one homeobox DNA binding domain belonging to the CUT homeobox family. SATB1 contributes to the initiation of X inactivation mediated by Xist RNA that occurs during embryogenesis and in lymphoma. SATB1 is organized into a cage-like network anchoring loops of heterochromatin and regulates gene expression through chromatin remodeling/HDAC (histone deacetylase complex), and transcription factors recruitment. SATB1 is predominantly expressed in thymus. Recent studies show that SATB1 is necessary for breast cancer cells to become metastatic.

Specificity

This antibody recognizes the C-terminus of SATB1.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human SATB1.

Application

Detect Special AT-rich sequence-binding protein 1 using this rabbit polyclonal antibody, Anti-SATB1 Antibody validated for use in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected SATB1 in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected SATB1 in human brain cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in A20 cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected SATB1 in 15 µg of A20 cell lysate.

Target description

~90 and ~98 kDa observed. Uniprot describes two isoforms at ~86 kDa and ~89 kDa Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A20 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinem Örnek et al.
Journal of cutaneous pathology, 51(3), 232-238 (2023-11-07)
Diagnosis of early mycosis fungoides (eMF) is challenging and often delayed as many of its clinical and histopathologic features may mimic various benign inflammatory dermatoses (BIDs). The products of the thymocyte selection-associated high mobility group box (TOX), twist family BHLH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service