Skip to Content
Merck
All Photos(2)

Key Documents

ABE472

Sigma-Aldrich

Anti-WAC Antibody, NT

from rabbit, purified by affinity chromatography

Synonym(s):

WW domain-containing adapter protein with coiled-coil

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€411.00

€411.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€411.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€411.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WAC(51322)

General description

WAC (WW domain-containing adapter protein with coiled-coil) functions as a linker between gene transcription and histone H2B monoubiquitination at ′Lys-120′ (H2BK120ub1). Wac interacts with the RNA polymerase II transcriptional machinery through its WW domain and with RNF20-RNF40 via its coiled-coil region. It has been shown to regulate H2BK120ub1, gene transcription, cell-cycle checkpoint activation in response to DNA damage, positively regulate amino acid starvation-induced autophagy and negatively regulate the ubiquitin proteasome pathway. In nuclear speckles it has been observed to colocalize with pre-mRNA processing complexes. This antibody recognizes the N-terminus of the WAC protein.

Immunogen

Recombinant protein corresponding to human WAC.

Application

Anti-WAC, N-terminus detects levels of WAC proteins & has been published & validated for use in WB, IP & ChIP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: 1 µg/mL from a representative lot detected WAC in 10 µg of HEK293 cell lysate.

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected WAC, N-terminus in 293T cell lysates stably transfected with SFB-RNF20 and in HCT116 cell lysates transfected with siRNA negative control (Zhang, F. and Yu, X. (2011). Mol Cell. 41(4):384-397.).

Chromatin Immunoprecipitation: A representative lot from an independent laboratory detected WAC, N-terminus in HCT116 cell lysates transfected with siRNA negative control, but demonstrated a loss of signal in HCT166 cell lysates transfected with siRNF20 (Zhang, F. and Yu, X. (2011). Mol Cell. 41(4):384-397.).

Quality

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected WAC, N-terminus in 10 µg of K562 cell lysate.

Target description

~80 kDa observed. Multiple isoforms are known to exist at ~70 kDa, ~65 kDa, ~60 kDa, and ~47 kDa Uncharacterized bands may be observed at ~52 kDa, ~75 kDa, and ~270 kDa in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
K562 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service