Skip to Content
Merck
All Photos(2)

Key Documents

ABE316

Sigma-Aldrich

Anti-ARID1B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

AT-rich interactive domain-containing protein 1B, ARID domain-containing protein 1B, BRG1-associated factor 250b, BAF250B, BRG1-binding protein hELD/OSA1, Osa homolog 2, hOsa2, p250R

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€497.00

€497.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€497.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€497.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARID1B(57492)

General description

ARID1B (AT rich interactive domain 1B), also known as BAF250B, p250R and hOsa2, is a nuclear protein and a member of the SWI/SNF-A chromatin-remodeling complex. It is involved in repression of select genes by chromatin remodeling and transcriptional activation. It non-specifically binds DNA and alters DNA nucleosome topology. The cause of mental retardation autosomal dominant type 12 (MRD12) are defects in ARID1B. High levels of ARID1B can be found in heart, skeletal muscle and kidney.

Immunogen

KLH-conjugated linear peptide corresponding to human ARID1B.

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated ARID1B from Raji cell nuclear extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-ARID1B Antibody is validated for use in Western Blotting, Immunoprecipitation for the detection of ARID1B.

Quality

Evaluated by Western Blot in Raji cell nuclear extract.

Western Blot Analysis: 1.0 µg/mL of this antibody detected ARID1B in 10 µg of Raji cell nuclear extract.

Target description

~250 kDa observed. Uncharacterized bands may be observed at ~16 kDa and ~32 kDa in some lysates.. Uniprot describes 4 isoforms produced by alternative splicing at ~236 kDa, ~237 kDa, ~241 kDa and ~162 kDa This antibody will recognize all isoforms.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service