Skip to Content
Merck
All Photos(1)

Key Documents

ABE2871

Sigma-Aldrich

Anti-phospho-Smad1 (Ser463 Antibody, Ser465)

from rabbit

Synonym(s):

Mothers against decapentaplegic homolog 1, MAD homolog 1, Mothers against DPP homolog 1, JV4-1, Mad-related protein 1, SMAD family member 1, hSMAD1, Transforming growth factor-beta-signaling protein 1, BSP-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer463 )

Gene Information

human ... SMAD1(4086)

General description

Mothers against decapentaplegic homolog 1 (UniProt: Q15797; also known as MAD homolog 1, Mothers against DPP homolog 1, JV4-1, Mad-related protein 1, SMAD family member 1, SMAD1, Smad1, hSMAD1, Transforming growth factor-beta-signaling protein 1, BSP-1) is encoded by the SMAD1 (also known as BSP1, MADH1, MADR1) gene (Gene ID: 4086) in human. Smad1 is a transcriptional modulator that is activated by BMP (bone morphogenetic protein) type 1 receptor kinase. Smad1 is a receptor-regulated Smad (R-Smad) that is found in a complex with Smad4 and YY1. Upon phosphorylation at the C-terminal by BMP type 1 receptor kinase it forms a trimer with another Smad1 and Smd4. The C-terminal phosphoserines of Smad1 are recognized by the Mad Homology 2 (MH2) domain of Smad4. The Smd1/Smad4 complex then translocates to the nucleus where Smad proteins bind to their cognate DNA binding sites with low affinity. This binding is further enhanced in the presence of transcriptional co-activators. Smad1 is ubiquitinated by Smad-specific E3 ubiquitin ligase SMURF1, leading to its degradation via proteasome Smad1 variants are associated with susceptibility to pulmonary hypertension that is characterized by plexiform lesions of proliferating endothelial cells in pulmonary arterioles.

Specificity

This rabbit polyclonal antibody detects Smad1 phosphorylated on Ser463 and Ser465. it targets an epitope within 8 amino acids surrounding Ser463 and Ser465.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 8 amino acids surrounding pSer463/pSer465 from the C-terminus region of human Smad1.

Application

Anti-phospho-Smad1 (Ser463, Ser465), Cat. No. ABE2871, is a highly specific rabbit polyclonal antibody that targets Mothers against decapentaplegic homolog 1 (Smad1) and has been tested in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected phospho-Smad1 (Ser463, Ser465) in constitutively active ALK6 version in 35S metabolic labeled COS cells (Persson, U., et. al. (1998). FEBS Lett. 434(1-2):83-7).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HepG2 cells stimulated with BMP7.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected phospho-Smad1 (Ser463, Ser465) in HepG2 cells stimulated with BMP7.

Target description

~57 kDa observed; 52.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.01% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service