Skip to Content
Merck
All Photos(3)

Key Documents

ABE2588-25UL

Sigma-Aldrich

Anti-H3Q5dopaminyl

Synonym(s):

Dopaminylated Histone H3|H3Q5dopaminylated

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€152.00

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€152.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€152.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

human, rat

technique(s)

dot blot: suitable
inhibition assay: suitable (peptide)
western blot: suitable

General description

Histone H3.1t (UniProt: Q16695; also known as H3/t, H3t, H3/g) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histone H3 has two main variants, H3.1 and H3.3, which show different genomic localization patterns in animals. The H3.1 and H3.3 complexes also possess distinct histone chaperones, CAF-1 and HIRA, which play important role in mediating DNA-synthesis-dependent and -independent nucleosome assembly. Histone H 3.1 is a core component of nucleosome that is present only in mammals. It is expressed during S phase, then its expression decreases significantly as cell division slows down during the process of differentiation. Histone H 3.1 expression is shown to be replication dependent. It serves as the canonical histone, which is incorporated during DNA replication. Histones are dynamic proteins that undergo multiple types of post-translational modifications, including acetylation, phosphorylation, methylation, and ubiquitination. Histone 3.1 is also shown to undergo serotonylation of glutamine at position 5 (Q5ser) in organisms that produce serotonin. It has been reported that Histone H3 can also undergo dopaminylation ((H3Q5dop), which can potentiate the binding of adjacent histone post-translational modification interacting proteins and play a vital role in dopaminergic neuronal transcription. This rabbit polyclonal antibody specifically binds to Histone H3 dopaminylated on glutamine 5. (Ref.: Lepack, AE., et al (2020). Science. 368(6487); 197-201).

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 with dopaminylated glutamine 5.

Application

Anti-H3Q5dopaminyl, Cat. No. ABE2588, is a rabbit polyclonal antibody that detects Histone H3 dopaminylated on glutamine 5 and is tested for use in Dot Blot, Peptide Inhibition Assay, and Western Blotting.
Western Blotting Analysis: A 1:200 dilution from a representative lot detected H3Q5dopaminyl in human ventral tegmental area (Courtesy of Ian Maze Lab; Icahn School of Medicine at Mount Sinai, New York).

Dot Blot Analysis: A representative lot de

Quality

Evaluated by Western Blotting with recombinant H3 dopaminyated on glutamine 5.

Western Blotting Analysis: A 1:500 dilution of this antibody detected recombinant Histone H3 dopaminylated on glutamine 5 with 2.5 mM dopamine, but not with one treated with transglutaminase 2 (0.5 mg).

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service