Skip to Content
Merck
All Photos(2)

Key Documents

ABE2585

Sigma-Aldrich

Anti-Mis12 Complex

from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€280.00

€280.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€280.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€280.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Mis12 complex is composed of four subunits, Protein MIS12 homolog, Polyamine-modulated factor 1, Kinetochore-associated protein DSN1 homolog, and Kinetochore-associated protein NSL1 homolog (UniProt: Q9H081, Q6P1K2, Q9H410, Q96IY1, respectively) that are encoded by genes known as MIS12 (Gene ID: 79003), PMF1 (Gene ID: 100527963), DSN1 (Gene ID: 79980), and NSL1 (also known as C1orf48, DC31, DC8, MIS14) (Gene ID: 25936) in human. The MIS12 complex is a protein interaction hub for outer kinetochore assembly. This complex acts as the primary microtubule-binding interface at kinetochores and provides a platform to recruit regulatory proteins. In human Mis12 complex subunits are shown to localize with centromere protein A (CENP-A) at inner kinetochores and internally to Ndc80 at outer kinetochores. Mis12 complex plays an essential role in chromosome segregation in vertebrates and contributes to mitotic kinetochore assembly. Reduced levels of Mis12 complex proteins are shown to result in chromosome alignment defects in both human and chicken cells, but spindle bipolarity is not disturbed. (Ref.: Kline SL et al (2006). J. Cell. Biol. 173(1): 9-17).

Specificity

This polyclonal antibody detects Mis12 complex in human. It will also detect individual subunits (Mis12, PMF1, Dsn1, Nsl1) in Western Blotting.

Immunogen

Epitope: unknown
Four subunits from human Mis12 complex.

Application

Anti-Mis12 Complex Antibody, Cat. No. ABE2585, is a highly specific rabbit polyclonal antibody that targets Mis12 complex and has been tested in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Immunocytochemistry Analysis: A 1:1,100 dilution from a representative lot detected Mis12 complex in HeLa cells.

Immunofluorescence Analysis: A representative lot detected Mis12 complex in HeLa cells (Rago, F., et. al. (2015). Curr Biol. 25(5):671-7).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in recombinant Mis12 complex.

Western Blotting Analysis: 2.2 µg/mL of this antibody detected Mis12 complex in 1 µL of recombinant Mis12 complex.

Target description

24.10, 23.34, 40.07, and 32.16 kDa calulated for Mis12, PMF1, DSN1, and NSL1, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS without azide and with 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service