Skip to Content
Merck
All Photos(2)

Key Documents

ABE238

Sigma-Aldrich

Anti-Katanin-like 1 (KL1) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Katanin p60 ATPase-containing subunit A-like 1, Katanin p60 subunit A-like 1, p60 katanin-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KATNAL1(84056)

General description

Katanin-like 1 (KL1) is a mitotic regulator that is very closely related to katanin. KL1 is thought to be involved in microtubule severing because increased expression of KL1 leads to total disassembly of microtubules. KL1 is seen to localize to the spindle poles during mitosis, not to the centrosome.

Immunogen

KLH-conjugated linear peptide corresponding to human KL1.

Application

Anti-Katanin-like 1 (KL1) Antibody is a Rabbit Polyclonal Antibody for detection of Katanin-like 1 (KL1) also known as Katanin p60 subunit A-like 1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot Analysis: A 1:1.000 dilution of this antibody detected KL1 on 10 µg of U2OS cell lysate.

Immunocytochemistry: A representative lot of this antibody was used for IC by an independent laboratory (Sonbuchner, T.M., et al. (2010). Cell Cycle. 9(12):2403-2411).

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:1.000 dilution of this antibody detected KL1 on 10 µg of HeLa cell lysate.

Target description

~55 kDa observed. Uncharacterized bands at ~210, 150, 63, and 45 may be observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service