Skip to Content
Merck
All Photos(1)

Key Documents

ABE1932

Sigma-Aldrich

Anti-IRX-5

from rabbit

Synonym(s):

Iroquois-class homeodomain protein IRX-5, Homeodomain protein IRXB2, Iroquois homeobox protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€295.00

€295.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€295.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€295.00


Estimated to ship on24 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Irx5(54352)

General description

Iroquois-class homeodomain protein IRX-5 (UniProt Q9JKQ4; also known as Homeodomain protein IRXB2, Iroquois homeobox protein 5) is encoded by the Irx5 (also known as Irxb2) gene (Gene ID 54352) in murine species. Iroquois homeobox (Irx) genes encode a family of homeodomain transcription factors that are conserved from worms to vertebrates. Six IRX genes are located in two clusters in human (IRXA and IRXB) and mouse (IrxA and IrxB) genomes. Irx3 and Irx5 of the IrxB cluster are expressed in the developing mouse limb bud and are essential for proximal and anterior skeletal formation. Irx3 and Irx5 are required to establish early anteroposterior (AP) polarity and specify the anterior progenitor population. Individual Irx genes are also important for certain postnatal cardiac functions in the developing mouse heart, including cardiac repolarization (Irx5) and rapid ventricular conduction (Irx3). Postnatal deletion of Irx3 and Irx5 in the myocardium leads to prolongation of atrioventricular conduction, due in part to activation of expression of the Na(+) channel protein Nav1.5. However, combined postnatal loss of Irx3 and Irx5 results in a restoration of the repolarization gradient that is altered in Irx5 mutant hearts, suggesting that postnatal Irx3 activity can be repressed by Irx5 in mice.

Specificity

This rabbit polyclonal antiserum targets an internal region within the C-terminal half of murine IRX-5/IRXB2.

Immunogen

His-tagged recombinant murine IRX-5/IRXB2 C-terminal fragment GST fusion protein (Chen, C.W., et al. (2005). Dev. Biol. 287(1):48-60).

Application

Research Category
Epigenetics & Nuclear Function
This rabbit polyclonal Anti-IRX-5 Antibody, Cat. No. ABE1932 detects levels of IRX-5, and has been published and validated for use in Western Blotting.
Western Blotting Analysis: A representative lot was affinity purified and detected an increased IRX-5/IRXB2 expression in postnatal P10 heart tissue lysates from miR-1-2-knockout mice (Zhao, Y., et al. (2007). Cell. 129(2):303-317).

Quality

Evaluated by Western Blotting in postnatal mouse heart tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antiserum detected IRX-5/IRXB2 in 10 µg of postnatal P10 mouse heart tissue lysate.

Target description

~54 kDa observed. 50.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service