Skip to Content
Merck
All Photos(3)

Key Documents

ABE1836

Sigma-Aldrich

Anti-SMARCAL1 Antibody

serum, from rabbit

Synonym(s):

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1, HepA-related protein, hHARP, Sucrose nonfermenting protein 2-like 1, SMARCAL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1 (UniProt Q9NZC9; also known as ATP-driven annealing helicase, HepA-related protein, hHARP, SMARCA-like protein 1, Sucrose nonfermenting protein 2-like 1) is encoded by the SMARCAL1 (also known as HARP, HHARP) gene (Gene ID 50485) in human. The annealing helicase SMARCAL1 is a replication stress response protein that maintains genome integrity at stalled replication forks during DNA replication. SMARCAL1 binds Replication protein A (RPA) via its RPA-binding motif, which is both necessary and sufficient to target SMARCAL1 to stalled replication forks. Although not necessary for SMARCAL1 enzymatic activity, RPA binding is critical for its cellular function. Ataxia-telangiectasia mutated (ATM), ATM and Rad3-related (ATR), and DNA-dependent protein kinase (DNA-PK) phosphorylate SMARCAL1 in response to replication stress. SMARCAL1-deficient cells are hypersensitive to replication stress agents and SMARCAL1 gene mutations cause Schimke immunoosseous dysplasia (SIOD).

Specificity

Reacts with both human and murine SMARCAL1. Expected to react with all four spliced isoforms of murine SMARCAL1.

Immunogen

Recombinant protein corresponding to the N-terminus of human SMARCAL1.

Application

Anti-SMARCAL1 Antibody, Cat. No. ABE1836, is a highly specific rabbit Polyclonal antibody, that targets SMARCAL1 and has been tested in Western Blotting, Immunoprecipitation.
Immunoprecipitation Analysis: 3.0 µL from a representative lot immunoprecipitated SMARCAL1 in HEK293T cell lysate (Data courtesy of Dr. David Cortez, Vanderbilt University).
Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected SMARCAL1 in siRNA transfected HEK293T cell lysate (Data courtesy of Dr. David Cortez, Vanderbilt University).
Western Blotting Analysis: A representative lot detected SMARCAL1 in U2OS cell lysate (Bansbach, C.E., et al. (2009). Genes Dev. 23(20):2405-2414).

Quality

Evaluated by Western Blotting in mouse myoblast C2C12 nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SMARCAL1 in 10 µg of mouse myoblast C2C12 nuclear extract.

Target description

~110 kDa observed. Human SMARCAL1 (105.9 kDa calculated). Mouse SMARCAL1 (100.8 kDa/isoform 1, 87.1 kDa/isoform 2, 63.2 kDa/isoform 3, 44.2 kDa/isoform 4 calculated). Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhe Yang et al.
Genes & development, 34(19-20), 1392-1405 (2020-09-05)
TRF1 facilitates the replication of telomeric DNA in part by recruiting the BLM helicase, which can resolve G-quadruplexes on the lagging-strand template. Lagging-strand telomeres lacking TRF1 or BLM form fragile telomeres-structures that resemble common fragile sites (CFSs)-but how they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service