Skip to Content
Merck
All Photos(2)

Key Documents

ABE1803

Sigma-Aldrich

Anti-Adenosylhomocysteinase (AHCY)

from rabbit, purified by affinity chromatography

Synonym(s):

AdoHcyase, S-adenosyl-L-homocysteine hydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€401.00

€401.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on07 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... AHCY(191)

General description

Adenosylhomocysteinase (UniProt: P23526; also known as EC: 3.3.1.1, AdoHcyase, S-adenosyl-L-homocysteine hydrolase) is encoded by the AHCY (also known as SAHH) gene (Gene ID: 191) in human. Adenosylhomocysteinase is a homotetrametic enzyme that catalyzes the reversible hydrolysis of S-adenosylhomocysteine to adenosine and L-homocysteine. Hence, it regulates the intracellular S-adenosylhomocysteine concentration, which is thought to be important for transmethylation reactions. Two isoforms of adenosylhomocysteinase have been reported that are produced by alternative splicing. It contains three nucleotide binding regions (aa 157-159; 222-227; and 299-301). Defects in AHCY gene cause hypermethioninemia that is characterized with failure to thrive, mental and motor retardation, facial dysmorphism with abnormal hair and teeth, and myocardiopathy.

Specificity

This rabbit polyclonal antibody detects Adenosylhomocysteinase in human. It targets and epitope within 12 amino acids from the C-terminus region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminus region of human Adenosylhomocysteinase.

Application

Anti-Adenosylhomocysteinase (AHCY) Antibody, Cat. No. ABE1803, is a highly specific rabbit polyclonal antibody that targets Adenosylhomocysteinase (AHCY) and has been tested for use in Immunohistochemistry (Paraffin), and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 2 µg/mL from a representative lot detected Adenosylhomocysteinase (AHCY) in 200 µg of human placental tissue lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Adenosylhomocysteinase (AHCY) in human kidney and human skin tissues.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Adenosylhomocysteinase (AHCY) in 200 µg of HeLa cell lysate.

Target description

~47 kDa observed; 47.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH 7.4, with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service