Skip to Content
Merck
All Photos(2)

Key Documents

ABE1454

Sigma-Aldrich

Anti-NR4A1 (Nur77)

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor subfamily 4 group A member 1, Early response protein NAK1, Nuclear hormone receptor NUR/77, Nur77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, ST-59, Testicular receptor 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€320.00

€320.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€320.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€320.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, monkey, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NR4A1(3164)

General description

Nuclear receptor subfamily 4 group A member 1 (UniProt: P22736; also known as Early response protein NAK1, Nuclear hormone receptor NUR/77, Nur77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, ST-59, Testicular receptor 3) is encoded by the NR4A1 (also known as GFRP1, HMR, NAK1) gene (Gene ID: 3164) in human. Nur77 is an orphan nuclear receptor that may act concomitantly with NURR1 in regulating the expression of delayed-early genes during liver regeneration. It is shown to be expressed in fetal muscle and in adult liver, brain, and thyroid and is reported to be induced by growth stimulating agents. It binds the NGFI-B response element (NBRE) 5′-AAAAGGTCA-3′. Its DNA binding region is localized to amino acids 264-339. It contains two zinc finger domains (aa 267-287 and 303-327). Nur77 is shown to heterodimerize with retinoid X receptor alpha and is actively transported from the nucleus to the mitochondria in cells undergoing apoptosis. It binds to Bcl-2 and triggers a conformational change that exposes the pro-apoptotic BH3 domain. This converts Bcl-2 from an anti-apoptotic to a pro-apoptotic factor. In several cancers the nuclear-mitochondrial translocation of Nur77/TR3 is defective, which prevents apoptosis. Three isoforms of Nur77 have been described that are produced by alternative splicing. It is acetylated by p300/CBP that increases its stability. It can also undergo phosphorylation at Serine 351 by RPS6KA1 and RPS6KA3 in response to mitogenic or stress stimuli.

Specificity

This rabbit polyclonal antibody detects Nuclear receptor subfamily 4 group A member 1 (NR4A1) in human, Monkey, and Murine cells It targets an epitope within 17 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Nuclear receptor subfamily 4 group A member 1 (NR4A1). The immunogen sequence is conserved in all 3 isoforms.

Application

Anti-NR4A1 (Nur77), Cat. No. ABE1454, is a highly specific rabbit polyclonal antibody that targets Nuclear receptor subfamily 4 group A member 1 and has been tested for use in Immunocytochemistry and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 2 µg/mL from a representative lot detected NR4A1 (Nur77) in 10 µg of COS7 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected NR4A1 (Nur77) in HeLa, A431, HUVEC, and NIH/3T3 cells.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected NR4A1 (Nur77) in 10 µg of HeLa cell lysate.

Target description

~64 kDa observed; 64.46 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service