Skip to Content
Merck
All Photos(2)

Key Documents

ABE1351

Sigma-Aldrich

Anti-GATA-6 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Transcription factor GATA-6, GATA-binding factor 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€324.00

€324.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€324.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€324.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GATA6(2627)

General description

GATA-6 is a developmental transcriptional activator important in terminal differentiation and proliferation of cells destined to be associated with the heart, gut and gut related tissues and organs. GATA-6 works in concert with SEMA3C and PLXNA2 transcription factors and regulates the expression of their target genes. GATA-6 is particularly important in heart development as a number of genetic mutations within GATA-6 give rise to defective syndromes involving the heart or the vessels of the heart. Interestingly, recent research into pulmonary hypertension disease in adults points to a critical role of GATA-6 expression in endothelial cells lining the blood vessels. Healthy blood vessels express GATA-6 very strongly but expression is lost during vascular injury and markedly down regulated in the endothelial cells found in diseased vessels. Because GATA-6 is a direct transcriptional regulator or important vascular proteins such as endothelin-1 receptor, endothelial nitric oxide synthase, and cytokines such as Fractalkine, GATA-6’s role in modulating the affects of pulmonary hypertension and other vascular syndromes is a subject of much research.

Specificity

Recognizes GATA-6

Immunogen

Epitope: Region between amino acid positions 250-500.
Set of 3 peptides designed against isoform 1 located between position 250-500). Note that peptide sequences used are also present in the isoform 2.

Application

Immunocytochemistry Analysis: A representative lot of this antibody has been reported to detect GATA-6 in HeLa, 293T, Capan-2, and SH-SY5Ycells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-GATA-6 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of GATA-6.

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: A 2 µg/mL dilution of this antibody detected GATA-6 in PC3cell lysate.

Target description

~ 62 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service