Skip to Content
Merck
All Photos(5)

Documents

ABE1348

Sigma-Aldrich

Anti-Histone H2A.Z Antibody (C-term)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2A.Z, H2A/z

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

ChIP: suitable (ChIP-seq)
ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2AFZ(3015)

General description

H2A.Z, also known as H2AFZ, is a variant of histone H2A that is highly conserved from yeast to humans. Like other histone variants, H2A.Z is encoded by a unique gene, resulting in a slightly divergent type of H2A with specialized functions. H2A.Z is involved in chromatin compaction by stabilizing the histone octamer within the nucleosome. It promotes gene expression through the creation of an open domain of chromatin that is more easily transcribed. H2A.Z is nonrandomly distributed throughout the genome, targeted primarily to pericentric heterochromatin, but excluded from the inactive X chromosome and the nucleolus. H2A.Z is associated with both euchromatin and facultative heterochromatin. DNA methylation can influence chromatin structure and effect gene silencing by excluding H2A.Z, and likewise, H2A.Z protects genes from DNA methylation. Thus, H2A.Z plays a role in regulating heterochromatin silencing and is involved in transcriptional control.

Specificity

Recognizes histone H2A.Z
wide range expected

Immunogen

KLH-conjugated linear peptide corresponding to region the near C-terminus of Human Histone H2A.Z.

Application

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody (1μg per reaction) was used for ChIP using chromatin from HeLa and K562 cells.

ChIP-seq: A representative lot of this antibody (0.5 μg per reaction) was used to immunoprecipitate K562 chromatin for ChIP-seq analysis.

Western Blotting Analysis: A 1:2,000 dilution of a represenative lot of this antibody detected histone H2A.Z in acid extracted HeLa cell lysate.

Immunocytochemistry: A 1:500 dilution from a representative lot detected histone H2A.Z in HeLa cells.
This Anti-Histone H2A.Z Antibody (C-term) is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and ChIP-seq and Immunocytochemistry and ELISA for the detection of Histone H2A.Z (C-term).

Quality

Evaluated by Western Blotting using recombinant histones

Western Blotting Analysis: A 1.5 µg/mL dilution of this antibody detected recombinant Histone H2A.Z, but not histones H1, H2A,H2A.X, H3, or H4.

Target description

~16 kDa observed

Physical form

Affinity purified rabbit polyclonal in PBS containing 0.05% azide and 0.05% ProClin 300.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlotta Zampieri et al.
Molecular oncology, 16(6), 1259-1271 (2021-12-18)
Somatic inactivation of p53 (TP53) mainly occurs as missense mutations that lead to the acquisition of neomorphic mutant protein forms. p53 mutants have been postulated to exert gain-of-function (GOF) effects, including promotion of metastasis and drug tolerance, which generally contribute
Anas Reda et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 49(8), 1285-1295 (2024-02-17)
Creating long-lasting memories requires learning-induced changes in gene expression, which are impacted by epigenetic modifications of DNA and associated histone proteins. Post-translational modifications (PTMs) of histones are key regulators of transcription, with different PTMs producing unique effects on gene activity
Gilda Stefanelli et al.
Cell reports, 36(7), 109551-109551 (2021-08-19)
Rapid removal of histone H2A.Z from neuronal chromatin is a key step in learning-induced gene expression and memory formation, but mechanisms underlying learning-induced H2A.Z removal are unclear. Anp32e was recently identified as an H2A.Z-specific histone chaperone that removes H2A.Z from
Firyal Ramzan et al.
Scientific reports, 10(1), 14331-14331 (2020-09-02)
Emerging evidence suggests that histone variants are novel epigenetic regulators of memory, whereby histone H2A.Z suppresses fear memory. However, it is not clear if altered fear memory can also modify risk for PTSD, and whether these effects differ in males
Klotilda Narkaj et al.
eNeuro, 5(5) (2018-11-13)
Memory formation is a protracted process that initially involves the hippocampus and becomes increasingly dependent on the cortex over time, but the mechanisms of this transfer are unclear. We recently showed that hippocampal depletion of the histone variant H2A.Z enhances

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service