Skip to Content
Merck
All Photos(2)

Key Documents

ABE1320

Sigma-Aldrich

Anti-ASXL2

from rabbit

Synonym(s):

Putative Polycomb group protein ASXL2, Additional sex combs-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ASXL2(55252)

General description

Putative Polycomb group protein ASXL2 (UniProt: Q76L83; also known as Additional sex combs-like protein 2) is encoded by the ASXL2 (also known as ASXH2, KIAA1685) gene (Gene ID: 55252) in human. ASXL2 is a putative Polycomb group (PcG) protein that acts by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. It is not required to initiate repression, but is essential to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. ASXL2 is shown to be involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). It also acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3. ASXL2 is reported to be essential for haematopoiesis and acts as a haploinsufficient tumor suppressor in leukemia. ASXL2 gene is one of most commonly mutated genes in AML patients bearing the AML1-ETO fusion oncoprotein and loss of ASXL2 is shown to promote AML1-ETO-mediated leukemogenesis. ASXL2 protein contains two LXXLL motifs (aa 303-307 and 887-891), which are required for an association with nuclear receptors. It also contains an atypical PHD-type zinc finger regions (aa 1397-1434). Two isoforms of ASXL2 are reported that are produced by alternative splicing. Mutation in ASXL2 gene have also been linked to the development of Shashi-Pena syndrome that is characterized by delayed psychomotor development, intellectual disability, and macrocephaly, and neonatal feeding difficulties. (Ref.: Micol, JB et al. (2017). Nat. Commun. DOI: 10.1038/ncomms15429).

Specificity

This rabbit polyclonal antibody detects Additional sex combs-like protein 2 (ASXL2) in human cells. It targets an epitope within 22 amino acids from the internal region.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 22 amino acids from the internal region of human Additional sex combs-like protein 2 (ASXL2).

Application

Anti-ASXL2, Cat. No. ABE1320, is a highly specific rabbit polyclonal antibody that targets Additional sex combs-like protein 2 and has been tested in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected ASXL2 in HeLa cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in Raji nuclear extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected ASXL2 in 10 µg of Raji nuclear extract.

Target description

~154 kDa observed; 153.82 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Langille et al.
Cancer discovery, 12(12), 2930-2953 (2022-09-16)
Systematically investigating the scores of genes mutated in cancer and discerning disease drivers from inconsequential bystanders is a prerequisite for precision medicine but remains challenging. Here, we developed a somatic CRISPR/Cas9 mutagenesis screen to study 215 recurrent "long-tail" breast cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service