Skip to Content
Merck
All Photos(3)

Key Documents

ABE1311

Sigma-Aldrich

Anti-SETD5

from rabbit, purified by affinity chromatography

Synonym(s):

SET domain-containing protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€317.00

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SETD5(55209)

General description

SET domain-containing protein 5 (UniProt: Q9C0A6; also known as Setd5) is encoded by the SETD5 (also known as KIAA1757) gene (Gene ID: 55209) in human. Setd5 protein is shown to be essential for mammalian development and is required for regulating germ cell-associated genes in embryonic stem cells. It interacts with Tbl1xr1 and Ctr9, which are components of 2 different transcriptional regulatory complexes (NcoR1 corepressor complex and Paf1 complex) in murine embryonic stem cells. Setd5-deficient embryonic stem cells display impaired cellular proliferation, increased apoptosis, defective cell cycle progression, a diminished ability to differentiate into cardiomyocytes, and greatly perturbed gene expression. Setd5-deficient mouse embryos exhibit severe defects in neural tube formation, somitogenesis and cardiac development, have aberrant vasculogenesis in embryos, yolk sacs, and placentas. They die between embryonic day 10.5 and 11.5. Three isoforms of Setd5 have been reported that are produced by alternative splicing. Its SET domain is localized to amino acids 269-390. Mutation in SETD5 gene has been linked to intellectual disability. (Ref.: Yu, SE., et al. (2017). Cell. Biochem. Funct. 35(5); 247-253; Osipovich, AB., et al. (2016). Development 143(24); 4595-4607).

Specificity

This rabbit polyclonal antibody detects SET domain containing protein 5 in human cells. It targets an epitope within 19 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human SET domain-containing protein 5.

Application

Anti-SETD5, Cat. No. ABE1311, is a highly specific rabbit polyclonal antibody that targets SET domain-containing protein 5 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected SETD5 in human cerebral cortex, human pancreas, and human testis tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected SETD5 in U2OS cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected SETD5 in 10 µg of HeLa nuclear extract.

Target description

~150 kDa observed; 158.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service