Skip to Content
Merck
All Photos(1)

Key Documents

ABE1082

Sigma-Aldrich

Anti-JARID1C Antibody

from rabbit

Synonym(s):

Lysine-specific demethylase 5C, Histone demethylase JARID1C, Jumonji/ARID domain-containing protein 1C, Protein SmcX, Protein Xe169

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€326.00

€326.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM5C(8242)

General description

JARID1C, also known as Jumonji/ARID domain-containing protein 1C, Protein SmcX, or Protein Xe169, and encoded by the gene KDM5C/DXS1272E/JARID1/SMCX/XE169, a histone demethylase that specifically demethylates ′Lys-4′ of histone H3, thereby playing a central role in histone code. JARID1C demethylates trimethylated and dimethylated but not monomethylated H3 ′Lys-4′. JARID1C does not demethylate histone H3 ′Lys-9′, H3 ′Lys-27′, H3 ′Lys-36′, H3 ′Lys-79′ or H4 ′Lys-20′. Additionally, JARID1C participates in transcriptional repression of neuronal genes by recruiting histone deacetylases and REST at neuron-restrictive silencer elements. JARID1C is localized to the nucleus. Defects in JARID1C are associated with Mental retardation, X-linked, syndromic, Claes-Jensen type (MRXSCJ) disorder. MRXSCJ patients have below average general intellectual functioning associated with impairments in adaptive behavior during development, and suffer mental retardation, progressive spastic paraplegia, seizures, and facial dysmorphism.

Immunogen

Linear peptide corresponding to human JARID1C.

Application

Anti-JARID1C Antibody detects level of JARID1C & has been published & validated for use in JARID1C.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected JARID1C in 10 µg of HEK293 cell lysate.

Target description

~175 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service