Skip to Content
Merck
All Photos(1)

Key Documents

ABE1043

Sigma-Aldrich

Anti-CAVIN-3/PRKCDBP Antibody

serum, from rabbit

Synonym(s):

Protein kinase C delta-binding protein, Cavin-3, Serum deprivation response factor-related gene product that binds to C-kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€278.00

€278.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€278.00


Estimated to ship on14 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

CAVIN-3/PRKCDBP, also known as Cavin-3/SRBC or Serum deprivation response factor-related gene product that binds to C-kinase, and encoded by the gene Prkcdbp/Srbc, is an adapter molecule that binds PKCdelta and is a member of the STICK (substrates that interact with C-kinase) superfamily of PKC-binding proteins. CAVIN-3 binds caveolin and localizes to the caveolae. CAVIN-3 remains associated with caveolin when caveolae bud to form vesicles (cavicles) that travel on microtubules to different regions of the cell. In the absence of CAVIN-3 intracellular cavicle traffic is markedly impaired. CAVIN-3 is also called SRBC for (sdr-related gene product that binds to c-kinase). Caveolae are a major membrane domain common to most cells. One of the defining features of this domain is the protein caveolin, and it is now know that CAVIN-3 is critical to caveolae function. CAVIN-3 is expressed in uterus, ovary, mammary and epithelial cells. Also expressed in brain, lung, heart, spleen, intestine, kidney, skeletal muscle and testis, but it is not found in liver. The protein appears to be up-regulated in serum starved conditions and growth arrest conditions suggesting a survival response.

Immunogen

GST-tagged recombinant protein corresponding to mouse CAVIN-3/PRKCDBP.

Application

Anti-CAVIN-3/PRKCDBP Antibody is a rabbit polyclonal antibody, validated for use in western blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected CAVIN-3/PRKCDBP in 10 µg of NIH/3T3 cell lysate.

Target description

~ 37 kDa observed. Uncharacterized band(s) may appear in some lysates. The calculated MW is 28 kDa However, this protein has been observed at ~34 kDa (Prof. U. Schibler, University of Geneva).

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maider Varela-Guruceaga et al.
Frontiers in physiology, 11, 565486-565486 (2020-12-17)
Repetitive complete or incomplete pharyngeal collapses are leading to chronic intermittent hypoxia (CIH), a hallmark feature of obstructive sleep apnea (OSA) syndrome responsible for many metabolic disorders. In humans, an association between OSA and insulin resistance has been found independently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service