Skip to Content
Merck
All Photos(1)

Key Documents

ABD133

Sigma-Aldrich

Anti-ACVR1B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Activin A receptor type IB, Activin receptor-like kinase 4, Activin receptor type 1B precursor, ACTR-IB, ACVRLK4, ALK4, ALK-4, Serine/threonine-protein kinase receptor R2, SKR2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€263.00

€263.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€263.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€263.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACVR1B(91)

General description

Activin receptor type-1B (ACVR1B) is a member of the protein kinase superfamily, and more specifically to the TKL Ser/Thr protein kinase family and TGFB receptor subfamily. ACVR1B, also known as activin receptor-like kinase 4 (ALK4), is a type I receptor for activin and plays major roles in cell differentiation, growth arrest and apoptosis. ACVR1B is known to form activin receptor complex with activin receptor type-2 (ACVR2A or ACVR2B). Within this receptor complex, type-2 receptors (ACVR2A and/or ACVR2B) act as a primary activin receptors whereas the type-1 receptors (including ACVR1B) act as downstream transducers of activin signals. Activins play role in many physiological and pathological processes including neuronal differentiation and neuronal survival, wound healing, extracellular matrix production, hair follicle development and cycling, FSH production, immunosuppression and carcinogenesis. ACVR1B is expressed highly in systemic sclerosis patient fibroblasts which suggests that the activin/ACRV1B signaling mechanism is involved in systemic sclerosis. ACVR1B is expressed in many tissues, most strongly in kidney, pancreas, brain, lung, and liver.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human ACVR1B.

Application

Anti-ACVR1B Antibody is a highly specific rabbit polyclonal antibody, that targets Activin receptor type-1 & has been tested in western blotting.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting/Peptide Inhibition Assay in human kidney tissue lysate.

Western Blotting/Peptide Inhibition Assay Analysis: 1 µg/mL of this antibody detected ACVR1B in 15 µg human kidney tissue lysate.

Target description

~70 kDa observed. Uniprot describes 5 isoforms ranging between ~52 kDa and ~62 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human kidney tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service