Skip to Content
Merck
All Photos(4)

Key Documents

ABC947

Sigma-Aldrich

Anti-Bif-1

from rabbit, purified by affinity chromatography

Synonym(s):

Endophilin-B1, Bax-interacting factor 1, SH3 domain-containing GRB2-like protein B1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€250.00

€250.00


Estimated to ship on03 June 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€250.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€250.00


Estimated to ship on03 June 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SH3GLB1(51100)

General description

Endophilin-B1 (UniProt: Q9Y371; also known as Bax-interacting factor 1, Bif-1, SH3 domain-containing GRB2-like protein B1) is encoded by the SH3GLB1 (KIAA0491; CGI-61) gene (Gene ID: 51100) in human.
Bif-1 is a member of the endophilin protein family that forms a complex with Beclin-1 through ultraviolet irradiation resistant-associated gene (UVRAG) and promotes the activation of the class III PI3 kinase, Vps34, in mammalian cells. It is highly expressed in heart, skeletal muscle, kidney, and placenta and is detected at lower levels in brain, colon, thymus, spleen, liver, small intestine, lung, and peripheral blood leukocytes. Loss of Bif-1 significantly suppresses PI3KC3 activation and the formation of autophagosome in both HeLa cells and mouse embryonic fibroblasts (MEFs). Bif-1 is reported to be involved in mitochondrial fission and coat protein complex I (COPI)-vesicle formation. Bif-1 contains a BAR domain at the N-terminal (aa 27-261) and a SH3 domain at the C-terminal (305-365), which is required for interaction with UVRAG. Bif-1 may be present as a homodimer or as a heterodimer with Endophilin-B2 (SH3GLB2). Bif-1 can be phosphorylated at Threonine 145 by Cdk5 and this phosphorylation is required for autophagy induction in starved neurons. Three isoforms of Bif-1 have been reported that are generated by alternative splicing. Isoform 1 acts as pro-apoptotic molecule in fibroblasts and is involved in caspase-independent apoptosis during nutrition starvation and in the regulation of autophagy. Isoform 2 has an anti-apoptotic role in neuronal cells and is involved in maintenance of mitochondrial morphology and promotes neuronal viability. (Ref.: Takahashi, Y et al (2009). Cell Death Differen. 16(7), 947-955).

Specificity

This rabbit polyclonal antibody detects Bax interacting facor 1 (Bif-1) in human cells. it targets an epitope within 15 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human Bax interacting factor 1 (Bif-1).

Application

Anti-Bif-1, Cat. No. ABC947, is a rabbit polyclonal antibody that detects Bax-interacting factor 1 (Bif-1) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Apoptosis & Cancer
Western Blotting Analysis: 2 µg/mL from a representative lot detected Bif-1 in 10 µg of Jurkat cells and human kidney lysates.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Bif-1 in human placenta, human skin, human small intestine, and human testis tissues.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Bif-1 in 10 µg of HEK293 cell lysate.

Target description

~52 kDa observed; 40.80 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service