Skip to Content
Merck
All Photos(1)

Key Documents

ABC940

Sigma-Aldrich

Anti-SNX18 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Sorting nexin-18, SH3 and PX domain-containing protein 3B, Sorting nexin-associated Golgi protein 1, SNX18

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€358.00

€358.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€358.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€358.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, canine

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SNX18(112574)

General description

Sorting nexin-18 (UniProt Q96RF0; also known as SH3 and PX domain-containing protein 3B, Sorting nexin-associated Golgi protein 1) is encoded by the SNX18 (also known as SNAG1, SH3PX2, SH3PXD3B) gene (Gene ID 112574) in human. SNX18, SNX33, and SNX9 constitute a subfamily of sorting nexin (SNX) proteins characterized by a Bin–Amphiphysin–Rvs (BAR) domain that allows dimerization. It also contributes to modulation and shaping of membrane curvature. It also has a Src-homology 3 (SH3) domain that allows interaction with a wide range of proline-rich PXXP motif containing proteins. In addition to mediating endocytosis and intracellular vesicle trafficking, SNX18, SNX33, and SNX9 are also essential for successful completion of the ingression and abscission stages of cytokinesis and for the delivery of a subset of vesicles to the intracellular bridge that connects the two nascent daughter cells during mitosis. GSK‐3 is reported to prevent FIP5-SNX18 interaction during metaphase and anaphase in Madin‐Darby canine kidney (MDCK) cells by catalyzing FIP5 Thr276 phosphorylation to ensure the fidelity and timing of FIP5‐endosome targeting to the AMIS during apical lumen formation.

Specificity

Expected to react with all spliced isoforms.

Immunogen

Recombinant protein corresponding to human SNX18.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-SNX18 Antibody is validated for use in Western Blotting, Immunoprecipitation, Immunocytochemistry for the detection of SNX18 .
Western Blotting Analysis: A representative lot detected endogenous SNX18 in MDCK and HeLa cell lysates (Willenborg, C., et al. (2011). J Cell Biol. 195(1):71-86).
Western Blotting Analysis: A representative lot detected endogenous SNX18 in MDCK cell lysates (Li, D., et al. (2014). EMBO J. 15(4):428-437).
Immunoprecipitation Analysis: A representative lot immunoprecipitated endogenous SNX18 in MDCK cell lysates (Willenborg, C., et al. (2011). J Cell Biol. 195(1):71-86).
Immunocytochemistry Analysis: A representative lot detected SNX18 in filter-grown, polarized MDCK cells, as well as polarized MDCK cells grown in 3D cultures (Willenborg, C., et al. (2011). J Cell Biol. 195(1):71-86).

Quality

Evaluated by Western Blotting in MDCK cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected SNX18 in 10 µg of MDCK cell lysate.

Target description

~73 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service