Skip to Content
Merck
All Photos(4)

Key Documents

ABC927

Sigma-Aldrich

Anti-LDHB Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

L-lactate dehydrogenase B chain (EC:1.1.1.27), LDH-B, LDH heart subunit, LDH-H, Renal carcinoma antigen NY-REN-46, LDHB

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€458.00

€458.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
100 μG
€458.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€458.00


Estimated to ship on22 April 2025Details


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), canine (based on 100% sequence homology), mouse (based on 100% sequence homology), rabbit (based on 100% sequence homology), bovine (based on 100% sequence homology), pig (based on 100% sequence homology), horse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LDHB(3945)

General description

L-lactate dehydrogenase B chain (EC 1.1.1.27; UniProt P07195; also known as Epididymis secretory protein Li 281, Lactate dehydrogenase H chain, LDH-B, LDH-H, LDH heart subunit, Renal carcinoma antigen NY-REN-46) is encoded by the LDHB (also known as LDHBD, TRG-5, HEL-S-281) gene (Gene ID 3945) in human. Originally described by Otto Warburg and now commonly known as the Warburg effect refers to the phenomenon that many tumor cells use aerobic glycolysis instead of mitochondrial respiration for energy production to support cell growth and proliferation. Enzymes involved in the complex network of metabolic pathways are therefore being studied as potential targets for cancer treatment. Lactate dehydrogenase (LDH) is a tetrameric enzyme composed of two different subunits, LDHA and LDHB, which can assemble into five different combinations (LDH1 is comprised of four LDHB subunits; LDH2 contains three LDHB and one LDHA subunit; LDH3 contains two LDHB and two LDHA subunits; LDH4 contains one LDHB and three LDHA subunits; LDH5 contains four LDHA subunits). LDHB is ubiquitously expressed and the predominant isoform in heart muscle, while LDHA is the predominant isoform in skeletal muscle and other highly glycolytic tissues. LDH catalyzes the reversible conversion of pyruvate into lactate. An upregulation of LDH therefore reduces the oxygen dependency and ensures an efficient anaerobic/glycolytic metabolism in tumor cells. Serum LDHB levels, for example, are reported to elevate in lung cancer patients and progressively increase with the clinical stage of non-small cell lung cancer (NSCLC).

Immunogen

KLH-conjugated linear peptide corresponding to human LDHB sequence from the C-terminal half.

Application

Detect LDHb using this rabbit polyclonal Anti-LDHB Antibody, Cat. No. ABC927, validated for use in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected LDHB in normal human prostate tissue, as well as human cancer cells in Non-Hodgkin lymphoma, kidney, colon, and lung tumor tissues.

Quality

Evaluated by Western Blotting in human PANC-1 cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected LDHB in 10 µg of human PANC-1 cell lysate.

Target description

~37 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maitham A Khajah et al.
Frontiers in pharmacology, 12, 747001-747001 (2021-11-09)
Background: Lactate dehydrogenase (LDH) plays an important role in cancer pathogenesis and enhanced expression/activity of this enzyme has been correlated with poor prognosis. In this study we determined the expression profile of LDH-A and B in normal as well as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service