Skip to Content
Merck
All Photos(1)

Key Documents

ABC514

Sigma-Aldrich

Anti-Menin/MEN1 Antibody

serum, from rabbit

Synonym(s):

Menin, Menin/MEN1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€421.00

€421.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on02 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

canine (based on 100% sequence homology), bovine (based on 100% sequence homology), human (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SCG2(7857)

General description

Menin (UniProt O00255) is a nuclear protein encoded by the MEN1 gene (also known as MEAI, SCG2) (Gene ID 4221) in human. MEN1 is a tumor suppressor gene frequently mutated in the familial multiple endocrine neoplasia type I (MEN-1) cancer syndrome characterized by tumors in multiple endocrine organs including the parathyroid, pancreatic islets, and the pituitary. Menin mediates cell proliferation and differentiation actions of transforming growth factor-beta (TGF-beta) ligand family members. Menin is reported to associate with histone methyltransferases (HMT) MLL1 (KMT2A) and MLL4 (KMT2B) to form menin-HMT complexes in both human and murine model systems. In addition, menin is also shown to be involved in DNA repair with FANCD2.

Specificity

Expected to react with all three spliced variants: Isoform 1 (Long), Isoform 2 (Short), and Isoform 3.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Menin/MEN1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Menin/MEN1 Antibody is validated for use in Western Blotting for the detection of Menin/MEN1 .
Western Blotting Analysis: A representative lot detected endogenous Menin in TGP61 nuclear extract, as well as exogenously expressed Menin in transfected NIH3T3 cells (Jin, S., et al. (2003). Cancer Res. 63(14):4204-4210).

Quality

Evaluated by Western Blotting in Tamoxifen treated MEF 545T and MEF 545D cell lysate.

Western Blotting Analysis: A 1:20,000 dilution of this antibody detected Menin/MEN1 in 10 µg of Tamoxifen treated MEF 545T and MEF 545D cell lysate.

Target description

~70 kDa observed

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Zeng et al.
Cancer research and treatment (2021-07-09)
Emerging evidence has shown that SKP1-cullin-1-F-box-protein (SCF) E3 ligases contribute to the pathogenesis of different cancers by mediating the ubiquitination and degradation of tumor suppressors. However, the functions of SCF E3 ligases in the pathogenesis of colorectal cancer (CRC) remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service