Skip to Content
Merck
All Photos(2)

Key Documents

ABC508

Sigma-Aldrich

Anti-BTG3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein BTG3, Abundant in neuroepithelium area protein, BTG family member 3, Protein Tob5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BTG3(10950)

General description

BTG3 is a tumor suppression protein because overexpression impairs serum-induced cell cycle progression from G0/G1 to the S phase in vitro. BTG3 is often down regulated in cancer cell lines, and thus BTG3 is a target for enhanced cancer therapy. In normal cells BTG3 inhibits cell proliferation, cell invasion, and angiogenesis, and also regulates cell-cycle progression and differentiation in a variety of cell types. Developmentally BTG3 is normally expressed primarily in the brain and CNS during development where is modulates cell proliferation. In the adult expression is high in ovary, testis, prostate, thymus and lung. It is not expressed strongly in many cancers.

Specificity

This antibodies immunogen is 94% identical in mouse and 92% identical in rat.

Immunogen

GST-tagged recombinant protein corresponding to human BTG3.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected BTG3 in human testis and human prostate tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-BTG3 Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of BTG3.

Quality

Evaluated by Western Blotting in IMR-90 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected BTG3 in 10 µg of IMR-90 cell lysate.

Target description

~37 kDa observed. Uncharacterized bands may be observed in some lysates. The calculated molecular weight of this protein is 29 kDa, but can be observed at ~37 kDa due to alternate splice variants.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service