Skip to Content
Merck
All Photos(2)

Key Documents

ABC482

Sigma-Aldrich

Anti-ADE2 Antibody/SAICAR synthetase

from rabbit, purified by affinity chromatography

Synonym(s):

Multifunctional protein ADE2, Phosphoribosylaminoimidazole-succinocarboxamide synthase, SAICAR synthetase, Phosphoribosylaminoimidazole carboxylase, AIR carboxylase, AIRC

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€287.00

€287.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€287.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€287.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

canine (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), porcine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), feline (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAICS(10606)

General description

Multifunctional protein ADE2, which is encoded by the gene name PAICS, ADE2, AIRC, PAIS, is a member of the SAICAR synthetase family and the AIR carboxylase family. Multifunctional protein ADE2 includes the following two domains: Phosphoribosylaminoimidazole-succinocarboxamide synthase (SAICAR synthetase), and Phosphoribosylaminoimidazole carboxylase (AIR carboxylase/ AIRC). ADE2 is a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. ADE2 is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for ADE2. ADE2/ PAICS (phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase) is a protein-coding gene. Diseases associated with ADE2 include androgen insensitivity syndrome, and pseudohermaphroditism, and among its related super-pathways are Metabolic pathways and Purine metabolism.

Immunogen

Epitope: SAICAR synthetase
KLH-conjugated linear peptide corresponding to human ADE2/SAICAR synthetase.

Application

This Anti-ADE2 antibody is validated for use in Western Blotting for the detection of ADE2.

Quality

Evaluated by Western Blotting in TF-1, K562, HL-60, THP-1, Jurkat, and MOLT-4 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected ADE2/SAICAR synthetase in 10 µg of TF-1, K562, HL-60, THP-1, Jurkat, and MOLT-4 cell lysate.

Target description

~47 kDa observed. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service