Skip to Content
Merck
All Photos(2)

Key Documents

ABC253

Sigma-Aldrich

Anti-Beclin-2

from rabbit

Synonym(s):

Beclin2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€401.00

€401.00


Estimated to ship on18 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€401.00


Estimated to ship on18 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Becn2(226720)

General description

Beclin-2 (UniProt: P0DM65) is encoded by Becn2 gene (Gene ID: 226720) in murine species. Beclin-2 is a member of the Atg6/Beclin family and is involved in 2 distinct lysosomal degradation pathways and acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. It regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. Beclin-2 family members are specific to mammals whereas Becln-1 subfamily members are present in more primitive vertebrates. Beclin-2 interacts with ATG14, AMBRA1, UVRAG and PIK3C3/VPS34 and this interaction is not disrupted by starvation. It is expressed in brain, skeletal muscle, placenta, thymus and uterus and expression is lower in liver, testis, stomach, and 17-day-old embryos. Disruption of Beclin-2 is shown to reduce embryonic viability and heterozygous knockout mice show defects in autophagy, increased levels of Cnr1 receptor, elevated food intake, and obesity and insulin resistance. (Ref.: He, C. et al. (2013). Cell 154, 1085-1099).

Specificity

This rabbit polyclonal antibody detects Becline-2 in murine cells. It targets an epitope within 14 amino acids from the N-terminal.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of murine Beclin-2.

Application

Anti-Beclin-2 Antibody, Cat. No. ABC253, is a rabbit polyclonal antibdoy that detects Beclin-2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Beclin-2 in human brain (thalamic region) and mouse stomach tissues.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in Beclin2 +/+ Mouse Brain (Wild-Type).

Western Blotting Analysis: 2 µg/mL of this antibody detected Beclin-2 in lysate from Beclin-2 +/+ Mouse Brain (Wild-type).

Target description

~50 kDa observed; 50.29 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon-Jin Kim et al.
Science advances, 7(8) (2021-02-21)
Drug abuse is a foremost public health problem. Cocaine is a widely abused drug worldwide that produces various reward-related behaviors. The mechanisms that underlie cocaine-induced disorders are unresolved, and effective treatments are lacking. Here, we found that an autophagy-related protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service